1.00665
Acetonitrile
≥99.8% (GC), suitable for GC/MS, SupraSolv®
Synonym(s):
ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide
About This Item
97 hPa ( 20 °C)
Recommended Products
Product Name
Acetonitrile, for gas chromatography MS SupraSolv®
vapor density
1.41 (vs air)
Quality Level
vapor pressure
72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)
product line
SupraSolv®
Assay
≥99.8% (GC)
form
liquid
autoignition temp.
524 °C
973 °F
expl. lim.
16 %
technique(s)
GC/MS: suitable
impurities
≤0.05% Water
evapn. residue
≤3.0 mg/L
color
APHA: ≤10
refractive index
n20/D 1.344 (lit.)
bp
81-82 °C (lit.)
mp
−45 °C (lit.)
transition temp
flash point 2 °C
density
0.786 g/mL at 25 °C (lit.)
format
neat
storage temp.
2-30°C
SMILES string
CC#N
InChI
1S/C2H3N/c1-2-3/h1H3
InChI key
WEVYAHXRMPXWCK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
- Thermodynamic Analysis of Eplerenone in 13 Pure Solvents at Temperatures from 283.15 to 323.15 K.: This study provides a comprehensive thermodynamic analysis of the solubility of Eplerenone in acetonitrile among other solvents, aiding in the optimization of pharmaceutical formulations and enhancing drug delivery systems (Liu et al., 2024).
- Preparation and characterization of stationary phase gradients on C8 liquid chromatography columns.: Detailing the use of acetonitrile in creating stationary phase gradients, this research supports advancements in chromatographic techniques for more efficient and selective separations in analytical chemistry (Cecil et al., 2024).
- Validation and application of an online extraction and liquid chromatography tandem mass spectrometry assay for the analysis of delamanid and its DM-6705 metabolite in human breast milk.: This article demonstrates the essential role of acetonitrile in the sample preparation and extraction for LC-MS/MS analysis, vital for drug monitoring and pharmacokinetic studies (Mkhize et al., 2024).
- Simultaneous quantification of 11 antiepileptic drugs using limited isotope-labeled internal standards in LC-MS/MS: An accuracy assessment.: Featuring acetonitrile′s application in the enhancement of LC-MS/MS methodologies, this study provides a robust framework for the accurate quantification of antiepileptic drugs, crucial for clinical pharmacology and therapeutic drug monitoring (Abady et al., 2024).
- Reductive Dynamic and Static Excited State Quenching of a Homoleptic Ruthenium Complex Bearing Aldehyde Groups.: This investigation highlights acetonitrile′s role in the study of photochemical reactions in ruthenium complexes, contributing to the understanding of light-induced processes in metal-organic frameworks (Dickenson et al., 2024).
Other Notes
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
35.6 °F - closed cup
Flash Point(C)
2.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service