Skip to Content
Merck
All Photos(5)

Key Documents

112933

Sigma-Aldrich

Acetanilide

99%

Synonym(s):

N-Phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H5
CAS Number:
Molecular Weight:
135.16
Beilstein:
606468
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39031308
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.65 (vs air)

Quality Level

vapor pressure

1 mmHg ( 114 °C)

Assay

99%

form

solid

autoignition temp.

1004 °F

bp

304 °C (lit.)

mp

113-115 °C (lit.)

functional group

amide

SMILES string

CC(=O)Nc1ccccc1

InChI

1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)

InChI key

FZERHIULMFGESH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetanilide undergoes palladium-catalyzed cross-coupling reaction to form ortho-acylacetanilide[1].

Application

Acetanilide is used as a EOF (electroosmotic flow) marker in the studies of affinity capillary electrophoresis for drug–protein binding[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

321.8 °F - closed cup

Flash Point(C)

161 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deia El-Hady et al.
Journal of pharmaceutical and biomedical analysis, 52(2), 232-241 (2010-01-19)
In order to achieve excellent precision in the estimation of binding constants by affinity capillary electrophoresis (ACE), electroosmotic flow (EOF) stability is the key parameter, especially when using proteins in binding assays. Appropriate rinsing protocols are mandatory. In our study
Zetty Azalea Sutirman et al.
International journal of biological macromolecules, 133, 1260-1267 (2019-05-03)
Grafting of crosslinked chitosan with monomer, N-vinyl-2-pyrrolidone, has been carried out to investigate its adsorption capacity toward Orange G (OG) from aqueous solutions. The adsorption performance of modified chitosan (cts(x)-g-PNVP) was examined and compared with that of the unmodified chitosan.
Gea H van der Lee et al.
Ecology, 102(3), e03275-e03275 (2020-12-23)
Changes in the ecological stoichiometry of primary producers may have considerable implications for energy and matter transfer in food webs. We hypothesized that nutrient enrichment shifts the trophic position of omnivores towards herbivory, as the nutritional quality of primary producers
Yinuo Wu et al.
Chemical communications (Cambridge, England), 49(7), 689-691 (2012-12-12)
A palladium-catalyzed cascade cross-coupling of acetanilide and toluene for the synthesis of ortho-acylacetanilide is described. Toluene derivatives can act as effective acyl precursors (upon sp(3)-C-H bond oxidation by a Pd/TBHP system) in the oxidative coupling between two C-H bonds. This
Pedro Henrique Cavalcanti Franco et al.
Magnetic resonance in chemistry : MRC, 58(1), 97-105 (2019-08-24)
Quantitative nuclear magnetic resonance (qNMR) is an analytical technique that offers numerous advantages in pharmaceutical applications including minimum sample preparation and rapid data collection times with no need for response factor corrections, being a powerful tool for assaying drug content

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service