Skip to Content
Merck
All Photos(3)

Key Documents

242381

Sigma-Aldrich

Benzoic acid

ACS reagent, ≥99.5%

Synonym(s):

Benzenecarboxylic acid, Carboxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOH
CAS Number:
Molecular Weight:
122.12
Beilstein:
636131
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39023903
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

4.21 (vs air)

vapor pressure

10 mmHg ( 132 °C)

Assay

≥99.5%

form

crystalline

autoignition temp.

1061 °F

packaging

poly bottle of 25, 100, 500 g
poly drum of 3 kg

impurities

MnO4- reducers, passes test
≤0.002% S compounds
≤0.005% CH3OH insol.

ign. residue

≤0.005%

bp

249 °C (lit.)

mp

121-125 °C (lit.)

solubility

water: soluble (2.9 g/l at 25 °C)

anion traces

chloride (Cl-): ≤0.005%

cation traces

heavy metals (as Pb): ≤5 ppm

functional group

carboxylic acid
phenyl

SMILES string

OC(=O)c1ccccc1

InChI

1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)

InChI key

WPYMKLBDIGXBTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoic acid is an organic aromatic monocarboxylic acid. It can be synthesized by the cobalt or manganese catalyzed atmospheric oxidation of toluene. Recently, benzoic acid has been prepared from toluene by employing TiO2 nanotubes electrode.
Benzoic acid reacts with hydrogenating reagents to afford hexahydrobenzoic acid. The thermal decomposition of the product in the presence of lime or alkali produces benzene and carbon dioxide.

Application

Benzoic acid has been used in the preparation of vials for the HPLC analysis of various polyamines in biological fluids, tissues and isolated/cultured cells.
It may be employed as an intermediate in the synthesis of the following:
  • paints
  • pigments
  • varnish
  • wetting agents
  • aroma compounds
  • benzoyl chloride
  • benzotrichloride
It may also be used to investigate the mechanism of complex addition reaction of hydroxyl radicals with various aromatic compounds.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaolai Dai et al.
Amino acids, 46(6), 1557-1564 (2014-03-19)
Polyamines (putrescine, spermine and spermidine) play a crucial role in the regulation of cell growth, differentiation, death and function. Accurate measurement of these substances is essential for studying their metabolism in cells. This protocol describes detailed procedures for sample preparation
Hydroxylation by electrochemically generated OH. radicals. Mono-and polyhydroxylation of benzoic acid: products and isomer distribution.
Oturan MA and Pinson J.
The Journal of Physical Chemistry, 99(38), 13948-13954 (1995)
Eagleson M.
Concise Encyclopedia Chemistry, 122-122 (1994)
Copper-mediated C-H/C-H biaryl coupling of benzoic acid derivatives and 1,3-azoles.
Mayuko Nishino et al.
Angewandte Chemie (International ed. in English), 52(16), 4457-4461 (2013-03-21)
Ly Dieu Tran et al.
Journal of the American Chemical Society, 134(44), 18237-18240 (2012-10-30)
An auxiliary-assisted, copper catalyzed or promoted sulfenylation of benzoic acid derivative β-C-H bonds and benzylamine derivative γ-C-H bonds has been developed. The method employs disulfide reagents, copper(II) acetate, and DMSO solvent at 90-130 °C. Application of this methodology to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service