Skip to Content
Merck
All Photos(1)

Key Documents

T-019

Supelco

(−)-11-nor-9-Carboxy-Δ9-THC solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O4
CAS Number:
Molecular Weight:
344.44
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

suitability

suitable for immunoassay

application(s)

cannabis testing
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCCCCC(C=C1O)=CC2=C1[C@@]3([H])[C@@](C(C)(C)O2)([H])CCC(C(O)=O)=C3

InChI

1S/C21H28O4/c1-4-5-6-7-13-10-17(22)19-15-12-14(20(23)24)8-9-16(15)21(2,3)25-18(19)11-13/h10-12,15-16,22H,4-9H2,1-3H3,(H,23,24)/t15-,16-/m1/s1

InChI key

YOVRGSHRZRJTLZ-HZPDHXFCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

11-nor-9-Carboxy-Δ9-THC is the inactive carboxylic form of 11-hydroxy-Δ9-tetrahydrocannabinol (Δ9-THC-OH), the primary oxidation bioactive metabolite of the main psychoactive component of cannabis, Δ9-Tetrahydrocannabinol (Δ9-THC).

Application

The Certified Spiking Solution® is suitable for (−)-11-nor-9-Carboxy-Δ9-THC testing by HPLC, GC/MS or LC-MS/MS methods in forensic analysis, clinical toxicology, urine drug testing, and Cannabis potency or impurity profiling testing applications. The Certified Reference Material (CRM) in solution can also be used as follows:
  • Analysis of Δ9-Tetrahydrocannabinol (Δ9-THC) and its three metabolites in plasma and urine samples by gas chromatography (GC) coupled to triple quadrupole mass spectrometer
  • Development of an automated sample preparation method based on solid phase extraction (SPE) to determine ∆9-tetrahydrocannabinol (THC) and its three primary metabolites in a variety of matrices, such as serum, saliva, and urine using liquid chromatography-mass spectrometry (LC-MS)
  • Simultaneous determination of cannabinoids and their metabolites in human serum samples by an atmospheric-pressure chemical ionization gas chromatography-tandem mass spectrometry (APGC-MS/MS)
  • Multi-analysis of cannabinoids and their metabolites by gas chromatography (GC) coupled to vacuum UV spectrometry
  • Liquid chromatography-tandem mass spectrometry (LC-MS/MS) method-based determination of 10 cannabinoids in oral fluid samples following their solid phase extraction (SPE)
  • Multi-residue analysis of 14 drugs of abuse from different categories by solid phase extraction (SPE) and LC-MS/MS

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia Coulter et al.
Journal of analytical toxicology, 36(6), 413-417 (2012-04-26)
This paper describes the determination of tetrahydrocannabinol (THC) and its metabolite, 11-nor-Δ⁹-tetrahydrocannabinol-9-carboxylic acid (THC-COOH) in oral fluid using solid-phase extraction and liquid chromatography with tandem mass spectral detection (LC-MS-MS) and its application to proficiency specimens. The method employs collection of
Vanessa de M Prata et al.
Analytical and bioanalytical chemistry, 403(2), 625-632 (2012-03-01)
A new procedure is described for the derivatization by silylation of 11-nor-Δ(9)-tetrahydrocannabinol-9-carboxylic acid (THCCOOH) present in urine, followed by analysis using gas chromatography-tandem mass spectrometry. A conventional procedure for derivatization of the analyte was evaluated using two types of experimental
Yannick Lecompte et al.
Journal of analytical toxicology, 36(8), 569-574 (2012-08-31)
The guidelines for screening of urinary cannabinoids require that all specimens testing positive should be confirmed by gas chromatography-mass spectrometry at a confirmatory test cutoff value of 15 ng/mL of 11-nor-9-carboxy-Δ9-tetrahydrocannabinol (THCCOOH). To assess the impact of lowering the confirmatory
Caroline Bylda et al.
Drug testing and analysis, 4(7-8), 668-674 (2012-03-01)
A fast and sensitive reference method for quantification of Δ(9) -tetrahydrocannabinol (THC) and its main metabolite 11-nor-9-carboxy-Δ(9) -tetrahydrocannabinol (THCCOOH) in oral fluid is described in this study. Samples were collected using an oral specimen collection device, followed by solid-phase extraction
Xiang He et al.
Analytical chemistry, 84(18), 7643-7647 (2012-08-21)
Oral fluid has been gaining more acceptance as the alternative matrix for forensic toxicology. Currently, Δ(9)-tetrahydrocannabinol (THC) is used as the primary target for detecting cannabis use in oral fluid. Meanwhile, THC carboxylic acid (THCA) in oral fluid is reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service