Skip to Content
Merck
All Photos(1)

Key Documents

W277304

Sigma-Aldrich

Neryl acetate

≥98%, stabilized, FCC, FG

Synonym(s):

cis-3,7-Dimethyl-2,6-octadien-1-yl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
FEMA Number:
2773
Beilstein:
1722814
EC Number:
Council of Europe no.:
2061
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.213
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥98%

may contain

<0.10% alpha-tocopherol, synthetic as stabilizer

composition

Contains IFRA and EU1223/2009 restricted Citral (Neral + Geranial) and Geraniol

refractive index

n20/D 1.460 (lit.)

bp

134 °C/25 mmHg (lit.)

density

0.91 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

geraniol, nerol, citral (neral+geranial)

Organoleptic

citrus; floral; fruity; rose; soapy; sweet

storage temp.

2-8°C

SMILES string

CC(=O)OC\C=C(\C)CC\C=C(/C)C

InChI

1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8-

InChI key

HIGQPQRQIQDZMP-FLIBITNWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Taste at 5 ppm

Other Notes

Natural occurrence: Orange juice, lemon, grapefruit, mandarin, lime and bergmot essential oils, black currant buds, grape, nutmeg, cocoa, tea, spice and buchu oil.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 28, 78-78 (2003)
Bio-degradation of acetates of geraniol, nerol & citronellol by P. incognita: isolation & identification of metabolites.
K M Madyastha et al.
Indian journal of biochemistry & biophysics, 20(3), 136-140 (1983-06-01)
Kye Chung Park et al.
Journal of insect physiology, 78, 33-46 (2015-05-06)
Phytophagous insects detect volatile compounds produced by host and non-host plants, using species-specific sets of olfactory receptor neurons (ORNs). To investigate the relationship between the range of host plants and the profile of ORNs, single sensillum recordings were carried out

Questions

  1. Quel est le pouvoir rotatoire de l'acetate de néryl

    1 answer
    1. This product does not contain any chiral centers, so the optical rotation is expected to be 0°. Note that the stabilizer is synthetic alpha-Tocopherol; while alpha-Tocopherol has three chiral centers, those centers are all racemic in the synthetic version. So the contribution of the stabilizer to the optical rotation would also be zero.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service