Skip to Content
Merck
All Photos(1)

Key Documents

W203203

Sigma-Aldrich

Allyl hexanoate

≥98%, FCC, FG

Synonym(s):

2-Propenyl hexanoate, Allyl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CO2CH2CH=CH2
CAS Number:
Molecular Weight:
156.22
FEMA Number:
2032
EC Number:
Council of Europe no.:
2181
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.244
NACRES:
NA.21
Organoleptic:
fruity; ethereal; pineapple; sweet
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.424 (lit.)

bp

75-76 °C/15 mmHg (lit.)

density

0.887 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; ethereal; pineapple; sweet

SMILES string

CCCCCC(=O)OCC=C

InChI

1S/C9H16O2/c1-3-5-6-7-9(10)11-8-4-2/h4H,2-3,5-8H2,1H3

InChI key

RCSBILYQLVXLJG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allyl hexanoate is a volatile flavoring compound with a pineapple flavor. It is naturally found in pineapple, baked potatoes and mushrooms. Allyl hexanoate is used as a flavoring agent in chewing gum, candies and baked goods.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fenaroli G
Fenaroli's Handbook of Flavor Ingredients, 18-18 (1971)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 91-91 (1997)
Yannai S
Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 923-923 (2012)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 91-91 (1997)
Robust dismissal of a claim on chemical sensitisation.
D Brahams
Lancet (London, England), 339(8788), 297-297 (1992-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service