Skip to Content
Merck
All Photos(1)

Documents

46015

Supelco

Neryl acetate

analytical standard

Synonym(s):

cis-3,7-Dimethyl-2,6-octadien-1-yl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
Beilstein:
1722814
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.460 (lit.)
n20/D 1.460

bp

134 °C/25 mmHg (lit.)

density

0.91 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)OC\C=C(\C)CC\C=C(/C)C

InChI

1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8-

InChI key

HIGQPQRQIQDZMP-FLIBITNWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Neryl acetate may be used as an analytical reference standard for the quantification of the analyte in essential oil samples using on-line high performance liquid chromatography coupled with high resolution gas chromatography and mass spectrometry (HPLC-HRGC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On-line high performance liquid chromatography coupled with high resolution gas chromatography and mass spectrometry (HPLC-HRGC-MS) for the analysis of complex mixtures containing highly volatile compounds
Mondello.L, et al.
Journal of Chromatography A, 39(9-10), 529-538 (1994)
Bio-degradation of acetates of geraniol, nerol & citronellol by P. incognita: isolation & identification of metabolites.
K M Madyastha et al.
Indian journal of biochemistry & biophysics, 20(3), 136-140 (1983-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service