Skip to Content
Merck
All Photos(1)

Key Documents

W277207

Sigma-Aldrich

Nerolidol

mixture of cis and trans, ≥97%, stabilized, FG

Synonym(s):

3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
222.37
FEMA Number:
2772
EC Number:
Council of Europe no.:
67
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.018
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.515

Assay

≥97%

contains

synthetic α-tocopherol as stabilizer

refractive index

n20/D 1.479 (lit.)

bp

114 °C/1 mmHg (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; floral; woody

SMILES string

C\C(C)=C/CC\C(C)=C\CCC(C)(O)C=C

InChI

1S/C15H26O/c1-6-15(5,16)12-8-11-14(4)10-7-9-13(2)3/h6,9,11,16H,1,7-8,10,12H2,2-5H3/b14-11+

InChI key

FQTLCLSUCSAZDY-SDNWHVSQSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Effects of Six Natural Compounds and Their Derivatives on the Control of Coccidiosis in Chickens.: This article discusses the use of natural compounds including nerolidol for controlling coccidiosis in poultry, indicating its applications in veterinary biochemistry and animal health (Hou et al., 2024).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S247-S250 (2008-07-22)
A toxicologic and dermatologic review of nerolidol when used as a fragrance ingredient is presented.
Shigeru Tamogami et al.
FEBS letters, 585(12), 1807-1813 (2011-04-23)
DMNT biosynthesis was proposed to proceed via (E)-nerolidol in plants a decade ago. However, (E)-nerolidol function as airborne signal/substrate for in-vivo biosynthesis of DMNT remains to be investigated and the regulation of DMNT production and emission is largely unknown. We
Fernanda Pículo et al.
Journal of applied toxicology : JAT, 31(7), 633-639 (2010-11-23)
Nerolidol is a sesquiterpenoid component of essential oil used as a flavor and aroma enhancer. It has also been studied as a topical skin penetration enhancer, and has inhibitory activities against S. aureus and E. coli, among other activities. The
S-Y Wang et al.
Insect molecular biology, 22(2), 172-182 (2013-01-09)
Olfaction plays an important role in insects' survival and reproduction. Odorant-binding proteins (OBPs) are considered to be one of the crucial proteins in the insect olfactory pathway. In this study, an antenna-specific OBP of the alfalfa plant bug, Adelphocoris lineolatus
Mahmoud AbouLaila et al.
Parasitology international, 59(2), 278-282 (2010-02-25)
Nerolidol is a sesquiterpene present in the essential oils of many plants, approved by the U.S. FDA as a food flavoring agent. Nerolidol interferes with the isoprenoid biosynthetic pathway in the apicoplast of P. falciparum. In the present study, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service