Skip to Content
Merck
All Photos(1)

Key Documents

166243

Sigma-Aldrich

cis-3,7-Dimethyl-2,6-octadien-1-yl acetate

98%

Synonym(s):

Neryl acetate, cis-3,7-Dimethyl-2,6-octadien-1-yl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
196.29
Beilstein:
1722814
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

impurities

1-2% geranyl acetate

refractive index

n20/D 1.460 (lit.)

bp

134 °C/25 mmHg (lit.)

density

0.91 g/mL at 25 °C (lit.)

functional group

ester

storage temp.

2-8°C

SMILES string

CC(=O)OC\C=C(\C)CC\C=C(/C)C

InChI

1S/C12H20O2/c1-10(2)6-5-7-11(3)8-9-14-12(4)13/h6,8H,5,7,9H2,1-4H3/b11-8-

InChI key

HIGQPQRQIQDZMP-FLIBITNWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-3,7-Dimethyl-2,6-octadien-1-yl acetate (Neryl acetate) is the major component of lavender volatile oils.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Robu et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 115(2), 584-589 (2011-08-30)
In this paper, we studied comparatively volatile fractions obtained by hydrodistillation from 5 samples of lavender flowers (L. angustifolia ssp. angustifolia, L. hybrida, L. angustifolia spp. pyrenaica, L. angustifolia spp. angustifolia cv. Munstead and cv. Hidcote blue) cultivated during 2009
Bio-degradation of acetates of geraniol, nerol & citronellol by P. incognita: isolation & identification of metabolites.
K M Madyastha et al.
Indian journal of biochemistry & biophysics, 20(3), 136-140 (1983-06-01)
Kye Chung Park et al.
Journal of insect physiology, 78, 33-46 (2015-05-06)
Phytophagous insects detect volatile compounds produced by host and non-host plants, using species-specific sets of olfactory receptor neurons (ORNs). To investigate the relationship between the range of host plants and the profile of ORNs, single sensillum recordings were carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service