Skip to Content
Merck
All Photos(1)

Key Documents

W266418

Sigma-Aldrich

(S)-(−)-Perillyl alcohol

≥95%, FG

Synonym(s):

p-Mentha-1,8-diene-7-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
FEMA Number:
2664
Council of Europe no.:
2024
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.060
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

optical activity

[α]20/D −88°, c = 1 in methanol

refractive index

n20/D 1.501 (lit.)

bp

119-121 °C/11 mmHg (lit.)

density

0.96 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green

SMILES string

CC(=C)[C@H]1CCC(CO)=CC1

InChI

1S/C10H16O/c1-8(2)10-5-3-9(7-11)4-6-10/h3,10-11H,1,4-7H2,2H3/t10-/m1/s1

InChI key

NDTYTMIUWGWIMO-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • CYP108N12 initiates p-cymene biodegradation in Rhodococcus globerulus.: This study explores the enzymatic breakdown pathways of monoterpenes, using (S)-(−)-Perillyl alcohol as a precursor, offering insights into microbial degradation processes that could be vital for bioremediation efforts or synthetic biology applications (Giang et al., 2022).

  • Orofacial antinociceptive effects of perillyl alcohol associated with codeine and its possible modes of action.: Research demonstrates the pain-relieving properties of (S)-(−)-Perillyl alcohol when combined with codeine, highlighting its potential for developing new analgesic formulations in dental and facial pain management (Limeira et al., 2022).

  • Orofacial antinociceptive activity of (S)-(-)-perillyl alcohol in mice: a randomized, controlled and triple-blind study.: This study underpins the effectiveness of (S)-(−)-Perillyl alcohol in reducing orofacial pain in a controlled experimental setup, providing a basis for further clinical trials in pain management (Tomaz-Morais et al., 2017).

  • In Vivo Anti-Tumor Activity and Toxicological Evaluations of Perillaldehyde 8,9-Epoxide, a Derivative of Perillyl Alcohol.: Highlights the anti-tumor properties of a novel derivative of (S)-(−)-Perillyl alcohol, suggesting its potential as a therapeutic agent in oncology, with comprehensive studies on its efficacy and safety (Andrade et al., 2016).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lars Duelund et al.
Journal of natural products, 75(2), 160-166 (2012-01-26)
The leaves of the annual plant Perilla frutescens are used widely as a spice and a preservative in Asian food as well as in traditional medicine. The active compounds in the leaves are the cyclic monoterpene limonene (1) and its
Ismail Kiran
Natural product communications, 6(12), 1805-1806 (2012-02-09)
S-(-)-Perillyl alcohol (p-mentha-1, 8-diene-7-ol) (1) (500 mg) was converted by Fusarium heterosporium ATCC 15625 over 10 days at 25 degrees C to a new metabolite, 1,2-dihydroxyperillyl alcohol (p-mentha-8-en-1,2,7-triol) (3) in a yield of 13% (70 mg). The structure of 3
Mohammad Farazuddin et al.
International journal of nanomedicine, 7, 35-47 (2012-01-26)
In the present study, a novel poly-lactic glycolic acid (PLGA)-based microparticle formulation of perillyl alcohol (POH) was prepared and characterized. Further, its efficacy was evaluated against di-methyl benzo anthracene-induced skin papilloma in Swiss albino mice. The characterization studies showed that
Juliana de Saldanha da Gama Fischer et al.
Journal of proteomics, 73(5), 1018-1027 (2010-01-20)
Perillyl alcohol (POH) is a naturally occurring terpene and a promising chemotherapeutic agent for glioblastoma multiform; yet, little is known about its molecular effects. Here we present results of a semi-quantitative proteomic analysis of A172 cells exposed to POH for
Juliana de Saldanha da Gama Fischer et al.
Journal of proteome research, 10(1), 153-160 (2010-09-03)
Glioblastoma multiform (GBM) is by far the most malignant glioma. We have introduced a new treatment for GBMs that comprises the inhalation of a naturally occurring terpene with chemotherapeutic properties known as perillyl alcohol (POH). Clinical trial results on recurrent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service