Skip to Content
Merck
All Photos(4)

Documents

D125806

Sigma-Aldrich

N,N-Diisopropylethylamine

ReagentPlus®, ≥99%

Synonym(s):

N-Ethyldiisopropylamine, DIPEA, Ethyldiisopropylamine, ‘Hünig’s base’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NC2H5
CAS Number:
Molecular Weight:
129.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

31 mmHg ( 37.7 °C)

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

127 °C (lit.)

mp

<−50 °C (lit.)

density

0.742 g/mL at 25 °C (lit.)

SMILES string

CCN(C(C)C)C(C)C

InChI

1S/C8H19N/c1-6-9(7(2)3)8(4)5/h7-8H,6H2,1-5H3

InChI key

JGFZNNIVVJXRND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Diisopropylethylamine, also known as Hünig′s base, is a sterically hindered amine. It is a non-nucleophilic base commonly employed for substitution reactions. It acts as an activator for chiral iridium N, P ligand complexes, which can be utilized in the hydrogenation of α, β-unsaturated nitriles. The influence of varying concentration of N,N-diisopropylethylamine on the synthesis of olvanil in the presence of lipase catalyst has been investigated.

Application

N,N-Diisopropylethylamine may be used in the synthesis of mannosylated ovalbumin peptides.
Proton scavenger used in peptide coupling, enolboration, Pd(0)-catalyzed alkoxycarbonylation of allyl phosphates and acetates, and as a catalyst in vinyl sulfone synthesis.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.1 °F

Flash Point(C)

9.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The Journal of Organic Chemistry, 58, 7162-7162 (1993)
The Journal of Organic Chemistry, 59, 695-695 (1994)
The Journal of Organic Chemistry, 58, 1538-1538 (1993)
Synthetic Communications, 23, 3073-3073 (1993)
Asymmetric Hydrogenation of α,β-Unsaturated Nitriles with Base-Activated Iridium N, P Ligand Complexes
Muller MA and Pfaltz A.
Angewandte Chemie (International Edition in English), 126(33), 8812-8815 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service