Skip to Content
Merck
All Photos(1)

Key Documents

77311

Supelco

(−)-Perillyl alcohol

analytical standard

Synonym(s):

(S)-4-Isopropenyl-1-cyclohexenylmethanol, (S)-p-Mentha-1,8-dien-7-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2501605
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D -88±2°, c = 1 in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.501

density

0.959 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(=C)[C@H]1CCC(CO)=CC1

InChI

1S/C10H16O/c1-8(2)10-5-3-9(7-11)4-6-10/h3,10-11H,1,4-7H2,2H3/t10-/m1/s1

InChI key

NDTYTMIUWGWIMO-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tabetha Sundin et al.
Molecular and cellular biochemistry, 375(1-2), 97-104 (2013-01-04)
We previously demonstrated in prostate cancer cells that a phytochemical-perillyl alcohol-and the mechanistic target of rapamycin (mTOR) inhibitor rapamycin rapidly attenuated telomerase activity. Protein levels of the telomerase catalytic subunit reverse transcriptase (hTERT) were diminished in the absence of an
Nonmelanoma skin cancer chemoprevention.
Renata Prado et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 37(11), 1566-1578 (2011-09-08)
Juliana de Saldanha da Gama Fischer et al.
Journal of experimental therapeutics & oncology, 7(4), 285-290 (2009-02-21)
Perillyl alcohol (POH) is a naturally occurring monoterpene with antiangiogenic and anti-tumoral properties. This chemotherapeutic agent has proven effectiveness in several clinical trials, including an ongoing phase I, comprising patients with recurrent glioblastoma multiform (GBM) under treatment with POH by
Donghak Kim et al.
Biotechnology letters, 31(9), 1427-1431 (2009-05-22)
The catalytic turnover of cytochrome P450( cam ) from Pseudomonas putida requires two auxiliary reduction partners, putidaredoxin (Pd) and putidaredoxin reductase (PdR). We report the functional expression in Escherichia coli of tricistronic constructs consisting of P450( cam ) encoded by
John L Clifford et al.
Cancer prevention research (Philadelphia, Pa.), 3(2), 132-135 (2010-01-28)
This perspective on Stratton et al. (beginning on p. 160), Kowalczyk et al. (beginning on p. 170), and Katiyar et al. (beginning on p. 179) highlights the common theme of translational investigation of natural substances and their molecular effects and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service