Skip to Content
Merck
All Photos(1)

Key Documents

M5905

Sigma-Aldrich

Mouse serum

Synonym(s):

Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
biological source:
mouse
sterility:
non-sterile
shipped in:
dry ice
Pricing and availability is not currently available.

biological source

mouse

Quality Level

sterility

non-sterile

contains

preservative

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Mouse serum acts as blocking agent in immunogold labeling[1], in vitro MBL-binding assays[2] and in the evaluation of commercial estradiol immunoassays.[3]
Mouse serum has been used as a:
  • blocking agent during the analysis of stem cell surface markers[4]
  • immunoprecipitation[5]
  • control in enzyme-linked immunosorbent assay[6]

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott M Goetz et al.
Biology open, 8(3) (2019-03-03)
When organisms possess chemical defenses, their predators may eventually evolve resistance to their toxins. Eastern indigo snakes (Drymarchon couperi; EIS) prey on pitvipers and are suspected to possess physiological resistance to their venom. In this study, we formally investigated this
D S Torres et al.
Biomaterials, 21(15), 1607-1619 (2000-07-08)
The contraction of connective tissue cells can play important roles in wound healing and pathological contractures. The effects of this contractile behavior on cell-seeded constructs for tissue engineering have not yet been investigated. The goal of this work was to
Malgorzata Gozdecka et al.
Nature genetics, 50(6), 883-894 (2018-05-08)
The histone H3 Lys27-specific demethylase UTX (or KDM6A) is targeted by loss-of-function mutations in multiple cancers. Here, we demonstrate that UTX suppresses myeloid leukemogenesis through noncatalytic functions, a property shared with its catalytically inactive Y-chromosome paralog, UTY (or KDM6C). In
Kristin M Scheible et al.
Clinical immunology (Orlando, Fla.), 161(2), 65-74 (2015-08-02)
Homeostatic T cell proliferation is more robust during human fetal development. In order to understand the relative effect of normal fetal homeostasis and perinatal exposures on CD8+ T cell behavior in PT infants, we characterized umbilical cord blood CD8+ T
Daniel J Haisenleder et al.
Endocrinology, 152(11), 4443-4447 (2011-09-22)
The University of Virginia Center for Research in Reproduction Ligand Core performed an evaluation of nine commercial estradiol (E2) immunoassays for use with mouse serum. The evaluation had two components. 1) Recovery Studies: a mouse pool was spiked with E2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service