Skip to Content
Merck
All Photos(3)

Documents

218391

Sigma-Aldrich

(S)-(−)-Perillyl alcohol

96%

Synonym(s):

p-Mentha-1,8-diene-7-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

optical activity

[α]22/D −88°, c = 1 in methanol

refractive index

n20/D 1.501 (lit.)

bp

119-121 °C/11 mmHg (lit.)

density

0.96 g/mL at 25 °C (lit.)

SMILES string

CC(=C)[C@H]1CCC(CO)=CC1

InChI

1S/C10H16O/c1-8(2)10-5-3-9(7-11)4-6-10/h3,10-11H,1,4-7H2,2H3/t10-/m1/s1

InChI key

NDTYTMIUWGWIMO-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(-)-Perillyl alcohol is a monoterpenoid compound found in the essential oils of cherries, lavender and spearmint. It shows potent anticancer activity.

Application

(S)-(−)-Perillyl alcohol (POH or 4-isopropenyl cyclohexene carbinol) can be used as a starting material in the synthesis of:
  • Perillyl alcohol neoglycoside derivatives as potential anti cancer agents.   
  • Amino-modified derivatives of (S)-perillyl alcohol as potent antiproliferative agents.        
  • Perillaldehyde 8,9-epoxide, a para-menthane class derivative as an in vivo anti-tumor agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Zi Hui et al.
Molecules (Basel, Switzerland), 19(5), 6671-6682 (2014-05-27)
Two series of amino-modified derivatives of (S)-perillyl alcohol were designed and synthesized using (S)-perillaldehyde as the starting material. These derivatives showed increased antiproliferative activity in human lung cancer A549 cells, human melanoma A375-S2 cells and human fibrosarcoma HT-1080 cells comparing
Nitin S Nandurkar et al.
Journal of medicinal chemistry, 57(17), 7478-7484 (2014-08-15)
A facile route to perillyl alcohol (POH) differential glycosylation and the corresponding synthesis of a set of 34 POH glycosides is reported. Subsequent in vitro studies revealed a sugar dependent antiproliferative activity and the inhibition of S6 ribosomal protein phosphorylation
Perillyl alcohol as a chemopreventive agent in N-nitrosomethylbenzylamine-induced rat esophageal tumorigenesis.
Liston BW, et al.
Cancer Research, 63(10), 2399-2403 (2003)
Metabolic engineering of Escherichia coli for limonene and perillyl alcohol production.
Alonso-Gutierrez J, et al.
Metabolic engineering, 19, 33-41 (2013)
Maria Antonieta Ferrara et al.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology], 44(4), 1075-1080 (2014-04-02)
Perillyl derivatives are increasingly important due to their flavouring and antimicrobial properties as well as their potential as anticancer agents. These terpenoid species, which are present in limited amounts in plants, may be obtained via bioconversion of selected monoterpene hydrocarbons.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service