Skip to Content
Merck
All Photos(3)

Documents

I3750

Sigma-Aldrich

3-Indoleacetic acid

98%

Synonym(s):

Heteroauxin, IAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
Beilstein:
143358
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

165-169 °C (lit.)

SMILES string

OC(=O)Cc1c[nH]c2ccccc12

InChI

1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)

InChI key

SEOVTRFCIGRIMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Indoleacetic acid (IAA) is a naturally occurring hydrophilic organic compound consists of an unsaturated aromatic ring that is commonly used in the synthesis of indolylated diarylmethanes, α-acetoxyl ketones, and Indolmethyl-Chromones via copper catalyzed coupling reactions.

Application

3-Indoleacetic acid (IAA) can be used as a building block to synthesize ,(±) harmacine using 4,4-diethoxybutan-1-amine via an acid-catalyzed acyl iminium ion cyclization reaction.
(±)-Harmacine is a vital step in the synthesis of a variety of indole alkaloids and dopamine/serotonin receptor ligands. IAA can also be used in the preparation of alkaloid (±)-19-hydroxyibogamine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper-Mediated Cyclization of o-Hydroxyaryl Enaminones with 3-Indoleacetic Acids toward the Synthesis of 3-Indolmethyl-Chromones
Kangmei W, et al.
The Journal of Organic Chemistry, 87, 9270-9281 (2022)
Effects of exogenous 3-indoleacetic acid and cadmium stress on the physiological and biochemical characteristics of Cinnamomum camphora
Jihai Z, et al.
Ecotoxicology and Environmental Safety, 191, 109998-109998 (2020)
A novel synthesis of (?)-harmacine and (?) 1, 2, 3, 4, 6, 7, 12, 12b-octahydroindole [2, 3-a] quinolizine
King Frank D
Journal of Heterocyclic Chemistry, 44(6), 1459-1463 (2007)
Microwave-assisted copper catalyzed decarboxylative reductive coupling of para-quinone methides with 3-indoleacetic acids: rapid access to polycyclic spiroindolequinone derivatives
Gui-Ting S, et al.
Inorganic chemistry frontiers, 10, 1512-1520 (2023)
Copper-Mediated Decarboxylative Coupling of 3-Indoleacetic Acids with Sulfoxonium Ylides for the Synthesis of alpha-Acetoxyl Ketones
Qiwen G, et al.
Organic Letters, 26, 5940-5945 (2024)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service