Z692867
BRAND® HandyStep® S manual repetitive pipette
Synonym(s):
Adjustable volume pipette
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.02
Recommended Products
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
BRAND 705110
volume measuring range
2 μL-5 mL
volume
5 mL
Related Categories
General description
Ideal for routine or research applications in diagnostics, molecular biology, environmental analysis, and many more.
HandyStep S and PD-tips work on the direct displacement principle. This offers the highest-precision dispensing of liquid media with high viscosity, high density, or high vapor pressure. Direct displacement permits contamination-free operation, since no aerosols are formed.
Includes:
HandyStep S, conformity-certified, with serial number, performance cerficate, operating manual, shelf/rack mount, 3 PD-Tips: 0.1 mL, 1 mL and 10 mL.
HandyStep S and PD-tips work on the direct displacement principle. This offers the highest-precision dispensing of liquid media with high viscosity, high density, or high vapor pressure. Direct displacement permits contamination-free operation, since no aerosols are formed.
- Excellent chemical resistance through innovative plastic materials
- Easy tip mounting - PD-tip is now simply inserted from below
- Dispensed volume range from 2 μL to 5 mL
- Up to 49 dispensing steps
- Lightweight and easy to handle
- Suitable for use with BRAND PD-tips, Encode tips, Repet tips, Combitips®
Includes:
HandyStep S, conformity-certified, with serial number, performance cerficate, operating manual, shelf/rack mount, 3 PD-Tips: 0.1 mL, 1 mL and 10 mL.
Features and Benefits
• Serial pipetting made easy• Works with BRAND PD-Tips and third-party tips• Robust, stable and durable
Other Notes
BR705110 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Combitips is a registered trademark of Eppendorf AG
HandyStep is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)