492817
Indole-3-acetic-2,2-d2 acid
98 atom % D, 98% (CP)
Synonym(s):
3-indoleacetic acid-α,α-d2, Deuterated indole-3-acetic acid, Heteroauxin-α,α-d2, IAA-α,α-d2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10D2H7NO2
CAS Number:
Molecular Weight:
177.20
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
isotopic purity
98 atom % D
Quality Level
Assay
98% (CP)
form
solid
technique(s)
mass spectrometry (MS): suitable
mp
165-169 °C (lit.)
mass shift
M+2
SMILES string
[2H]C([2H])(C(O)=O)c1c[nH]c2ccccc12
InChI
1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)/i5D2
InChI key
SEOVTRFCIGRIMH-BFWBPSQCSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Guizhi Feng et al.
Plant biotechnology journal, 19(7), 1337-1353 (2021-01-21)
Citrus fruit has a unique structure with soft leathery peel and pulp containing vascular bundles and several segments with many juice sacs. The function and morphology of each fruit tissue are different. Therefore, analysis at the organ-wide or mixed-tissue level
Hiroki Inahashi et al.
Physiologia plantarum, 164(2), 216-225 (2018-02-16)
Auxin flow is important for different root developmental processes such as root formation, emergence, elongation and gravitropism. However, the detailed information about the mechanisms regulating the auxin flow is less well understood in rice. We characterized the auxin transport-related mutants
Kathryn C Fitzgerald et al.
Cell reports. Medicine, 2(10), 100424-100424 (2021-11-11)
The circulating metabolome provides unique insights into multiple sclerosis (MS) pathophysiology, but existing studies are relatively small or characterized limited metabolites. We test for differences in the metabolome between people with MS (PwMS; n = 637 samples) and healthy controls (HC;
Lijie Xuan et al.
Plant, cell & environment, 43(3), 675-691 (2020-01-01)
Leaf trichomes protect against various biotic and abiotic stresses in plants. However, there is little knowledge about this trait in oilseed rape (Brassica napus). Here, we demonstrated that hairy leaves were less attractive to Plutella xylostella larvae than glabrous leaves.
Hong Wang et al.
Scientific reports, 10(1), 3745-3745 (2020-03-01)
During mung bean post-germination seedling growth, various metabolic and physiological changes occurred, leading to the improvement of its nutritional values. Here, transcriptomic and metabolomic analyses of mung bean samples from 6-hour, 3-day and 6-day after imbibition (6-HAI, 3-DAI, and 6-DAI)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service