Skip to Content
Merck
All Photos(2)

Documents

I2273

Sigma-Aldrich

3-Indoleacrylic acid

≥98% (HPLC), BioReagent

Synonym(s):

3-(3-Indolyl)acrylic acid, IAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO2
CAS Number:
Molecular Weight:
187.19
Beilstein:
6317
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

product line

BioReagent

Assay

≥98% (HPLC)

form

powder

mol wt

187.19  g/mol

storage temp.

room temp

SMILES string

OC(=O)\C=C\c1c[nH]c2ccccc12

InChI

1S/C11H9NO2/c13-11(14)6-5-8-7-12-10-4-2-1-3-9(8)10/h1-7,12H,(H,13,14)/b6-5+

InChI key

PLVPPLCLBIEYEA-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Indoleacrylic acid (IAA) is a chemical used for the induction of gene transcription. This expression system requires that the gene of interest is cloned in plasmids containing the trpE promoter.

Application

3-Indoleacrylic acid has been used in the induction of ribokinase expression in E. coli.

Biochem/physiol Actions

3-Indoleacrylic acid (IAA) is metabolized from tryptophan and this conversion is carried out by gut microflora. 3-Indoleacrylic acid (IAA) is a chemical used for the induction of gene transcription. The IAA induced expression system requires the gene of interest cloned in plasmids containing the trp promoter. It promotes the barrier functionality of the intestinal epithelial cells. IAA also favors anti-inflammatory response and is regarded as a potential therapeutic to treat inflammatory bowel disease (IBD). It is also implicated in a variety of cellular processes including apoptosis, autophagy, cell proliferation and migration. In tumor cells, IAA is also reported to hinder tryptophan metabolism.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indoleacrylic acid produced by commensal peptostreptococcus species suppresses inflammation
Wlodarska M, et al.
Cell host & microbe, 22(1), 25-37 (2017)
Identification and functional analyses of differentially expressed metabolites in early stage endometrial carcinoma
Shi K, et al.
Cancer Science, 109(4), 1032-1043 (2018)
Effective induction of tryptophan promoter under glucose starvation
MIZUTANI S, et al.
J. Chem. Eng. Jpn., 21(6), 658-660 (1988)
High-expression vectors with multiple cloning sites for construction of trpE fusion genes: pATH vectors.
T J Koerner et al.
Methods in enzymology, 194, 477-490 (1991-01-01)
Qiting Zhang et al.
International journal of molecular sciences, 22(11) (2021-06-03)
Epigenetic therapy using histone deacetylase (HDAC) inhibitors has become an attractive project in new drug development. However, DNA methylation and histone acetylation are important epigenetic ways to regulate the occurrence and development of leukemia. Given previous studies, N-(2-aminophenyl)benzamide acridine (8a)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service