GF45431647
Selenium
lump, 3 mm max. lump size, weight 50 g, high purity 99.999%
Synonym(s):
SE006110
About This Item
Recommended Products
description
gray
Assay
99.999%
form
lumps
manufacturer/tradename
Goodfellow 454-316-47
resistivity
1.2 μΩ-cm, 0°C
weight
50 g
max. lump size
3 mm
bp
684.9 °C (lit.)
mp
217 °C (lit.)
density
4.81 g/mL at 25 °C (lit.)
SMILES string
[Se]
InChI
1S/Se
InChI key
BUGBHKTXTAQXES-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT RE 2
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service