Skip to Content
Merck
All Photos(1)

Key Documents

324930

Sigma-Aldrich

Zinc

powder, <150 μm, 99.995% trace metals basis

Synonym(s):

Zinc metal powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 487 °C)

Assay

99.995% trace metals basis

form

powder

resistivity

5.8 μΩ-cm, 20°C

particle size

<150 μm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Related Categories

General description

Zinc powder is a fine, powdery substance made up of small particles (<150 micrometers) of the element zinc. Zinc powder is grey or silvered-colored and may have a metallic sheen. Because of the high-purity of this zinc powder, with very small concentration of trace metal impurities, it is suitable for the production of electronic components and semiconductor materials, for use as a catalyst in chemical reactions, and for research and testing.

Application

High purity zinc powder, with a particle size of less than 150 μm and a 99.995% trace metals basis, is a valuable material in several industrial and academic applications that require high purity levels. Due to its purity level, this zinc powder is specifically suitable for use in the production of semiconductors, where it serves as a dopant material in the manufacture of zinc oxide films. The high purity level ensures that it does not introduce impurities that could affect the performance of the semiconductor. High purity zinc powder is also essential in the production of metal powders using powder metallurgy techniques. It is mixed with other metal powders to produce alloys such as brass and bronze powders, which are commonly used in the manufacturing of automotive parts, industrial machinery, and electrical components. The high purity level ensures that the final product has the desired mechanical properties and does not introduce any impurities. In addition, high purity zinc powder is used as a reducing agent in various chemical reactions, such as the production of organic compounds and pharmaceuticals. The high purity level ensures that the final product is free from any contaminants that could impact its quality or effectiveness.

Features and Benefits

Our 99.995% pure zinc powder is useful in the electronics and chemical industries where purity matters most.
✔ Consistent quality
✔ Ultra high purity
✔ Available to scale up: bulk and pilot scale

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 5

1 of 5

Formation, cathodoluminescence and field emission of ZnO quantum dots attached on oxygen plasma activated carbon nanotubes
Proceedings, 361(2), 28-34 (2013)
Effects of dietary zinc on the growth, digestive enzyme activities, muscle biochemical compositions, and antioxidant status of the giant freshwater prawn Macrobrachium rosenbergii
Muralisankar T, et al.
Aquaculture (Amsterdam, Netherlands), 448, 98-104 (2015)
Application of zinc sealed tube graphitization on sub-milligram samples using EnvironMICADAS
Rinyu L, et al.
Nuclear Instruments and Methods in Physics Research, 361(2), 406-413 (2015)
Ultra small-mass graphitization by sealed tube zinc reduction method for AMS 14 C measurements
Xu X, et al.
Radiocarbon, 55(2), 608-616 (2013)
K(8)Tl(10)Zn: A Zintl Phase Containing the Zinc-Centered Thallium Polyanion Tl(10)Zn(8)(-).
Zhen-Chao Dong et al.
Inorganic chemistry, 36(16), 3559-3561 (1997-07-30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service