Skip to Content
Merck

Skip To

209651

Selenium

powder, −100 mesh, ≥99.5% trace metals basis

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
Se
CAS Number:
Molecular Weight:
78.96
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12161600
EC Number:
231-957-4
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

gray

Quality Segment

assay

≥99.5% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: selenium

resistivity

1.2 μΩ-cm, 0°C

particle size

−100 mesh

bp

684.9 °C (lit.)

mp

217 °C (lit.)

density

4.81 g/mL at 25 °C (lit.)

SMILES string

[Se]

InChI

1S/Se

InChI key

BUGBHKTXTAQXES-UHFFFAOYSA-N

General description

Selenium powder is a fine, powdery substance made up of small particles of the element selenium, often produced by the reduction of selenium oxide. Selenium powder is grey or silvered-colored and may have a metallic sheen that may appear reddish in color. This product is passed through a 100-mesh sieve, indicating that the particles are small and have a diameter of around 150 micrometers or less, and ensuring the powder is fine and easy-to-handle.

Application

Se is used in the synthesis of CdSe, which is consequetively used to modulate the photoresponse of quantum dot solar cells by varying the size and shape of semiconductor nanostructures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
204307GF33939849GF73071514
description

powder, −100 mesh, ≥99.5% trace metals basis

description

pellets, <5 mm particle size, ≥99.999% trace metals basis

description

powder, max. particle size 250 micron, weight 200 g, purity 99.95%

description

powder, 250 max. part. size (micron), purity 99.95%, weight 100 g

reaction suitability

reagent type: catalyst
core: selenium

reaction suitability

core: selenium

reaction suitability

-

reaction suitability

-

assay

≥99.5% trace metals basis

assay

≥99.999% trace metals basis

assay

99.95%

assay

99.95%

form

powder

form

pellets

form

powder

form

powder

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

-

bp

684.9 °C (lit.)

bp

684.9 °C (lit.)

bp

684.9 °C (lit.)

bp

684.9 °C (lit.)

density

4.81 g/mL at 25 °C (lit.)

density

4.81 g/mL at 25 °C (lit.)

density

4.81 g/mL at 25 °C (lit.)

density

4.81 g/mL at 25 °C (lit.)


Still not finding the right product?

Explore all of our products under Selenium


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters