Skip to Content
Merck
All Photos(1)

Key Documents

356808

Sigma-Aldrich

Iron

foil, thickness 0.1 mm, ≥99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Fe
CAS Number:
Molecular Weight:
55.85
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

≥99.9% trace metals basis

form

foil

reaction suitability

reagent type: catalyst
core: iron

resistivity

9.71 μΩ-cm

thickness

0.1 mm

bp

2750 °C (lit.)

mp

1535 °C (lit.)

density

7.86 g/mL at 25 °C (lit.)

SMILES string

[Fe]

InChI

1S/Fe

InChI key

XEEYBQQBJWHFJM-UHFFFAOYSA-N

General description

Iron foils may be used as a substrate for growing graphene by chemical vapor deposition (CVD).[1] Fragmentation of iron foils, using high power laser shock was investigated for engineering and astrophysical applications.[2]

Quantity

  • 50×50 mm (approximately 2 g)
  • 100×100 mm (approximately 8 g)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

High-power laser shock-induced dynamic fragmentation of iron foils.
Morard G, et al.
Physical Review. B, Condensed Matter and Materials Physics, 82(17), 174102-174102 (2010)
Synthesis of large-area, few-layer graphene on iron foil by chemical vapor deposition
Xue Y, et al.
Nano Research, 4(12), 1208-1214 (2011)
Elzbieta Krzesiek et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 34(203), 289-292 (2013-07-31)
Anemia is a frequent symptom of diseases of alimentary tract, also in children. Among others, inflammatory bowel disease, celiac disease and Helicobacter pylori are most often complicated by anemia. Not infrequently these disorders are accompanied by more than one type
Iron fortification and malaria risk in children.
Andrew M Prentice et al.
JAMA, 310(9), 914-915 (2013-09-05)
Shaun D Wong et al.
Nature, 499(7458), 320-323 (2013-07-23)
Mononuclear non-haem iron (NHFe) enzymes catalyse a broad range of oxidative reactions, including halogenation, hydroxylation, ring closure, desaturation and aromatic ring cleavage reactions. They are involved in a number of biological processes, including phenylalanine metabolism, the production of neurotransmitters, the

Questions

  1. I am reaching out to inquire about the manufacturing process of your Fe foil. Specifically, I would like to know whether it is produced through rolling, an electrolytic process, or another method.

    1 answer
    1. This foil product is cold rolled from thicker stock.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service