Skip to Content
Merck
All Photos(2)

Key Documents

GF73071514

Selenium

powder, 250 max. part. size (micron), purity 99.95%, weight 100 g

Synonym(s):

SE006010

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Se
CAS Number:
Molecular Weight:
78.96
MDL number:
UNSPSC Code:
12141910
PubChem Substance ID:
NACRES:
NA.23

description

gray

Assay

99.95%

form

powder

manufacturer/tradename

Goodfellow 730-715-14

resistivity

1.2 μΩ-cm, 0°C

weight

100 g

max. part. size

250 (micron)

bp

684.9 °C (lit.)

mp

217 °C (lit.)

density

4.81 g/mL at 25 °C (lit.)

SMILES string

[Se]

InChI

1S/Se

InChI key

BUGBHKTXTAQXES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sameer S Udhane et al.
Scientific reports, 7(1), 8652-8652 (2017-08-19)
Metformin is an antidiabetic drug, which inhibits mitochondrial respiratory-chain-complex I and thereby seems to affect the cellular metabolism in many ways. It is also used for the treatment of the polycystic ovary syndrome (PCOS), the most common endocrine disorder in
Jana Malikova et al.
The Journal of steroid biochemistry and molecular biology, 174, 192-200 (2017-09-13)
Abiraterone is an inhibitor of CYP17A1 which is used for the treatment of castration resistant prostate cancer. Abiraterone is known to inhibit several drug metabolizing cytochrome P450 enzymes including CYP1A2, CYP2D6, CYP2C8, CYP2C9, CYP2C19, CYP3A4 and CYP3A5, but its effects
Vincenza Barbato et al.
Fertility and sterility, 110(7), 1356-1366 (2018-12-07)
To study whether slush nitrogen (SN) vs. liquid nitrogen (LN) vitrification affects human ovarian tissue gene expression and preserves follicle health during extended in vitro culture. Randomized experimental study. University research laboratory. Ovarian biopsies collected by laparoscopic surgery from patients with
Patricia Rodríguez Castaño et al.
International journal of molecular sciences, 20(18) (2019-09-20)
Turmeric, a popular ingredient in the cuisine of many Asian countries, comes from the roots of the Curcuma longa and is known for its use in Chinese and Ayurvedic medicine. Turmeric is rich in curcuminoids, including curcumin, demethoxycurcumin, and bisdemethoxycurcumin.
L Dworak et al.
Nanoscale, 10(4), 2162-2169 (2018-01-13)
CdTe core and CdTe/CdSe core/shell quantum dots (QD) are investigated with steady state and time-resolved spectroscopic methods. The coating of the CdTe core with a 0.7 nm thick CdSe shell shifts the lowest exciton absorption band to the red by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service