Skip to Content
Merck
All Photos(1)

Key Documents

909831

Sigma-Aldrich

Oleylamine

Synonym(s):

cis-1-Amino-9-octadecene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CH2NH2
CAS Number:
Molecular Weight:
267.49
Beilstein:
1723960
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.23

vapor pressure

8 mmHg ( 135 °C)

form

liquid

refractive index

n20/D 1.4596 (lit.)

bp

348-350 °C (lit.)

mp

18-26 °C (lit.)

density

0.813 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCN

InChI

1S/C18H37N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10H,2-8,11-19H2,1H3/b10-9-

InChI key

QGLWBTPVKHMVHM-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oleyamine is widely used as a surfactant and reaction medium for shape control, colloidal stabilization, and surface functionalization of a variety of nanomaterials, including magnetic , metal, chalcogenide, rare earth, and quantum dot nanomaterial systems.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

309.2 °F - closed cup

Flash Point(C)

154 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleylamine in nanoparticle synthesis.
Mourdikousdis S, et al.
Chemistry of Materials, 25, 1465-1476 (2013)
Jiang Tang et al.
Nature materials, 10(10), 765-771 (2011-09-20)
Colloidal-quantum-dot (CQD) optoelectronics offer a compelling combination of solution processing and spectral tunability through quantum size effects. So far, CQD solar cells have relied on the use of organic ligands to passivate the surface of the semiconductor nanoparticles. Although inorganic
Synthesis and Characterization of Hydrophobic, Organically-Soluble Gold Nanocrystals Functionalized with Primary Amines.
Leff D, et al.
Langmuir, 12(20), 4723-4730 (1996)
A Simple Large-Scale Synthesis of Nearly Monodisperse Gold and Silver Nanoparticles with Adjustable Sizes and with Exchangeable Surfactants.
Hiramatsu H, et al.
Chemistry of Materials, 16(13), 2509-2511 (2004)
Copper Selenide Nanocrystals for Photothermal Therapy.
Hessel C M, et al.
Nano Letters, 11(6), 2560-2566 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service