Skip to Content
Merck
All Photos(1)

Documents

211141

Sigma-Aldrich

Lead(II) bromide

≥98%

Synonym(s):

Dibromolead, Lead dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbBr2
CAS Number:
Molecular Weight:
367.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98%

form

powder

reaction suitability

reagent type: catalyst
core: lead

bp

892 °C (lit.)

mp

371 °C (lit.)

solubility

ethanol: insoluble(lit.)

density

6.66 g/mL at 25 °C (lit.)

SMILES string

Br[PbH2]Br

InChI

1S/2BrH.Pb/h2*1H;/q;;+2/p-2

InChI key

ZASWJUOMEGBQCQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Lead(II) bromide (PbBr2) can be used in the fabrication of nanoscale quasi-2D layered perovskites, which are potentially utilized as light-emitting materials. It can also be used for the synthesis of deep blue fluorescent lead bromide perovskite microdisks. These microdisks can be used as direct bandgap semiconductors for light-emitting diodes (LEDs).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly luminescent nanoscale quasi-2D layered lead bromide perovskites with tunable emissions
Yuan Z, et al.
Chemical Communications (Cambridge, England), 52(20), 3887-3890 (2016)
A facile one-pot synthesis of deep blue luminescent lead bromide perovskite microdisks
Yuan Z, et al.
Chemical Communications (Cambridge, England), 51(91), 16385-16388 (2015)
A N Hamir et al.
Australian veterinary journal, 57(9), 401-406 (1981-09-01)
Eight-month-old dogs maintained on a high-fat-low-calcium diet were administered a mixture of lead chloride, lead bromide and lead sulphate for prolonged periods at 4 different dose levels. Dogs on high levels of leads showed marked weight loss and gastrointestinal symptoms
Hongying Duan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(7), 1447-1451 (2004-05-19)
In our experiments, it was observed that adding bromide to Pb2+ solution of N,N'-dimethylformamide (DMF), the highly emissive cluster Pb4Br11(3-) can be formed and the fluorescence intensity of the formed cluster is proportional to the concentration of Pb2+, based on
A O Maslat et al.
Journal of trace elements and electrolytes in health and disease, 3(4), 187-191 (1989-12-01)
The mutagenicity of lead (II) bromide (a combustion product of the gasoline additives lead (IV) tetraethyl and 1,2-dibromoethane) was investigated using various strains of bacteria. Taking prodigiosin (the red pigment) production as a marker, lead (II) bromide was found to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service