Skip to Content
Merck
All Photos(2)

Key Documents

325163

Sigma-Aldrich

Dodecylamine

≥99%

Synonym(s):

1-Aminododecane, Laurylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11NH2
CAS Number:
Molecular Weight:
185.35
Beilstein:
1633576
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

64 mmHg ( 170 °C)

Quality Level

Assay

≥99%

form

solid

bp

247-249 °C (lit.)

mp

27-29 °C (lit.)

solubility

water: soluble 3.5 g/L at 25 °C

density

0.806 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCN

InChI

1S/C12H27N/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-13H2,1H3

InChI key

JRBPAEWTRLWTQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Intercalation of dodecylamine into the layer space of kaolinite was investigated.[1] Dodecylamine was investigated as inhibitor of mild steel hydrochloric acid corrosion.[2]

Application

Dodecylamine was used in preparation of novel surfactant copper(II) complexes.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral - STOT SE 3

Target Organs

Gastrointestinal tract,Liver,Immune system, Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuai Zhang et al.
Journal of colloid and interface science, 430, 345-350 (2014-06-30)
Dodecylamine was successfully intercalated into the layer space of kaolinite by utilizing the methanol treated kaolinite-dimethyl sulfoxide (DMSO) intercalation complex as an intermediate. The basal spacing of kaolinite, measured by X-ray diffraction (XRD), increased from 0.72 nm to 4.29 nm
Substitutional inhibition mechanism of mild steel hydrochloric acid corrosion by hexylamine and dodecylamine.
Bastidas JM, et al.
J. Appl. Electrochem., 30(10), 1173-1177 (2000)
Darya V Radziuk et al.
Small (Weinheim an der Bergstrasse, Germany), 6(4), 545-553 (2010-01-29)
Alloyed gold/silver nanoparticles with a core/shell structure are produced from preformed gold and silver nanoparticles during ultrasonic treatment at different intensities in water and in the presence of surface-active species. Preformed gold nanoparticles with an average diameter of 15 +
Jozef Adamcik et al.
Langmuir : the ACS journal of surfaces and colloids, 25(5), 3159-3162 (2009-05-14)
It is shown that temperature-controlled ordered layers of dodecylamine, self-assembled on highly oriented pyrolytic graphite (HOPG), are an appropriate substrate for aligning individual DNA molecules. High resolution atomic force microscopy (AFM) permits visualization of the lamellar structure of dodecylamine on
Tukayi Kudanga et al.
Journal of biotechnology, 149(1-2), 81-87 (2010-07-06)
Enzymatic processes provide new perspectives for modification of lignocellulose materials. In the current study, laccase catalyzed coupling of long chain alkylamines to lignin model molecules and lignocellulose was investigated. Up to two molecules of dodecylamine (DA) and dihexylamine (DHA) were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service