Skip to Content
Merck
All Photos(1)

Key Documents

HT-OA100

SAFC

Oleylamine

≥98% (primary amine)

Synonym(s):

cis-1-Amino-9-octadecene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CH2NH2
CAS Number:
Molecular Weight:
267.49
Beilstein:
1723960
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor pressure

8 mmHg ( 135 °C)

Assay

≥98% (primary amine)

refractive index

n20/D 1.4596 (lit.)

bp

348-350 °C (lit.)

mp

18-26 °C (lit.)

density

0.813 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCN

InChI

1S/C18H37N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10H,2-8,11-19H2,1H3/b10-9-

InChI key

QGLWBTPVKHMVHM-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

309.2 °F - closed cup

Flash Point(C)

154 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ze Sun et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-10)
Magnesium hydride (MgH2) has been considered as a potential material for storing hydrogen, but its practical application is still hindered by the kinetic and thermodynamic obstacles. Herein, Mn-based catalysts (MnCl2 and Mn) are adopted and doped into MgH2 to improve
Zhicheng Zhang et al.
Science advances, 7(13) (2021-03-26)
Metallic nanostructures are commonly densely packed into a few packing variants with slightly different atomic packing factors. The structural aspects and physicochemical properties related with the vacancies in such nanostructures are rarely explored because of lack of an effective way
Cong-Min Fan et al.
Nanoscale, 7(7), 3247-3253 (2015-01-27)
The orthorhombic polymorph of Cu2ZnGeS4 (CZGS) is a metastable wurtzite-derived phase that can only be prepared in the bulk form by extensive heating at high temperatures (≥790 °C) when using the conventional solid-state reaction route. By employing a facile solution-based
Oleksandr Voznyy et al.
ACS nano, 13(10), 11122-11128 (2019-09-21)
Colloidal quantum dots (CQDs) allow broad tuning of the bandgap across the visible and near-infrared spectral regions. Recent advances in applying CQDs in light sensing, photovoltaics, and light emission have heightened interest in achieving further synthetic improvements. In particular, improving
Jongsik Park et al.
ACS nano, 12(8), 7996-8005 (2018-08-15)
Post-synthetic transformation of nanoparticles has received great attention, because this approach can provide an unusual route to elaborately composition-controlled nanostructures while maintaining the overall structure of the template. In principle, anisotropic heteronanoparticles of semiconductor materials can be synthesized via localized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service