Skip to Content
Merck
All Photos(2)

Key Documents

900599

Sigma-Aldrich

(4-(Aminomethyl)phenyl)(4-(prop-2-yn-1-yloxy)phenyl)methanone

95%

Synonym(s):

Amino benzophenone alkyne, Probe building block

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15NO2
Molecular Weight:
265.31
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

Application

(4-(Aminomethyl)phenyl)(4-(prop-2-yn-1-yloxy)phenyl)methanone is used for chemical probe synthesis, this trifunctional building block contains a light-activated benzophenone, alkyne tag, and amine synthetic handle. When appended to a ligand or pharmacophore through its amine linker, this building block allows for UV light-induced covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service