Skip to Content
Merck
All Photos(1)

Key Documents

764213

Sigma-Aldrich

Biotin-PEG4-alkyne

for copper catalyzed click labeling

Synonym(s):

Polyethylene glycol, Acetylene-PEG4-biotin conjugate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H35N3O6S
CAS Number:
Molecular Weight:
457.58
MDL number:
UNSPSC Code:
12161502
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

form

solid

reaction suitability

reaction type: click chemistry

mp

55-64 °C

storage temp.

−20°C

SMILES string

O=C(NCCOCCOCCOCCOCC#C)CCCC[C@@H](SC1)[C@@]2([H])[C@]1([H])NC(N2)=O

InChI

1S/C21H35N3O6S/c1-2-8-27-10-12-29-14-15-30-13-11-28-9-7-22-19(25)6-4-3-5-18-20-17(16-31-18)23-21(26)24-20/h1,17-18,20H,3-16H2,(H,22,25)(H2,23,24,26)/t17-,18-,20-/m1/s1

InChI key

SKMJWNZZFUDLKQ-QWFCFKBJSA-N

Application

Biotin-PEG4-alkyne may be used for the modification of 4-azidophenylalanine (AzPhe) silk fibroin via bioorthogonal azide–alkyne cycloaddition reaction for developing photopatternable protein material.[1]
Biotinylation reagent for labeling azide containing molecules or biomolecules using copper-catalyzed 1,3 dipolar cycloaddition click chemistry. The alkyne group reacts with azides to form a stable triazole linkage, facilitating the introduction of biotin into your azide modified system of interest.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masanobu Nagano et al.
Bioorganic & medicinal chemistry, 25(21), 5952-5961 (2017-10-11)
Vaccination is a reliable method of prophylaxis and a crucial measure for public health. However, the majority of vaccines cannot be administered orally due to their degradation in the harsh gut environment or inability to cross the GI tract. In
Anna Rutkowska et al.
ACS chemical biology, 11(9), 2541-2550 (2016-07-08)
Late stage failures of candidate drug molecules are frequently caused by off-target effects or inefficient target engagement in vivo. In order to address these fundamental challenges in drug discovery, we developed a modular probe strategy based on bioorthogonal chemistry that
Wei Wang et al.
Biochemistry, 56(30), 3889-3893 (2017-07-07)
The diverse gut microbial communities are crucial for host health. How the interactions between microbial communities and between host and microbes influence the host, however, is not well understood. To facilitate gut microbiota research, selective imaging of specific groups of
Maheshika S K Wanigasekara et al.
ACS omega, 3(10), 14229-14235 (2019-08-29)
Modification of arginine residues using dicarbonyl compounds is a common method to identify functional or reactive arginine residues in proteins. Arginine undergoes several kinds of posttranslational modifications in these functional residues. Identifying these reactive residues confidently in a protein or
Nrf1 can be processed and activated in a proteasome-independent manner.
Vangala JR, et al.
Current Biology, 26(18), R834-R835 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service