900615
(4-Hydroxyphenyl)(4-(prop-2-yn-1-yloxy)phenyl)methanone
≥95%
Synonym(s):
Hydroxyl benzophenone alkyne, Probe building block
About This Item
Recommended Products
Quality Level
Assay
≥95%
form
solid
reaction suitability
reaction type: click chemistry
storage temp.
−20°C
SMILES string
O(CC#C)c1ccc(cc1)C(=O)c2ccc(cc2)O
InChI
1S/C16H12O3/c1-2-11-19-15-9-5-13(6-10-15)16(18)12-3-7-14(17)8-4-12/h1,3-10,17H,11H2
InChI key
TZKIHDJSIXVMIQ-UHFFFAOYSA-N
Related Categories
Application
Other Notes
Technology Spotlight: Trifuctional Probe Building Blocks
A library approach to rapidly discover photoaffinity probes of the mRNA decapping scavenger enzyme DcpS
Design, synthesis and biological evaluation of photoaffinity probes of antiangiogenic homoisoflavonoids
Clickable Photoaffinity Ligands for Metabotropic Glutamate Receptor 5 Based on Select Acetylenic Negative Allosteric Modulators
γ-Secretase Modulator (GSM) Photoaffinity Probes Reveal Distinct Allosteric Binding Sites on Presenilin
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service