900602
4-(4-(Prop-2-yn-1-yloxy)benzoyl)benzoic acid
≥95%
Synonym(s):
Carboxyl benzophenone alkyne, Probe building block
About This Item
Recommended Products
Quality Level
Assay
≥95%
form
solid
reaction suitability
reaction type: click chemistry
reagent type: cross-linking reagent
functional group
alkyne
carboxylic acid
storage temp.
−20°C
SMILES string
O=C(C1=CC=C(OCC#C)C=C1)C2=CC=C(C(O)=O)C=C2
InChI
1S/C17H12O4/c1-2-11-21-15-9-7-13(8-10-15)16(18)12-3-5-14(6-4-12)17(19)20/h1,3-10H,11H2,(H,19,20)
InChI key
URDMKFAPEKSQDV-UHFFFAOYSA-N
Related Categories
Application
Other Notes
Technology Spotlight: Trifuctional Probe Building Blocks
A library approach to rapidly discover photoaffinity probes of the mRNA decapping scavenger enzyme DcpS
Synthesis and Evaluation of New Thiodigalactoside-Based Chemical Probes to Label Galectin-3
Discovery of tumor-specific irreversible inhibitors of stearoyl CoA desaturase
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service