Skip to Content
Merck
All Photos(2)

Documents

809993

Sigma-Aldrich

2-Propynyl methanesulfonate

battery grade, ≥99.5%, acid <200 ppm, H2O <100 ppm

Synonym(s):

2-Propinyl methanesulfonate, Methanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3S
CAS Number:
Molecular Weight:
134.15
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

Quality Level

description

Acid content: <200 ppm
Moisture: <100 ppm

Assay

≥99.5%

form

liquid

impurities

≤100 ppm H2O
≤200 ppm acid

bp

167 °C (lit.)

mp

17-19 °C (lit.)

density

1.481 g/cm3

application(s)

battery manufacturing

SMILES string

CS(OCC#C)(=O)=O

InChI

1S/C4H6O3S/c1-3-4-7-8(2,5)6/h1H,4H2,2H3

InChI key

OWAHJGWVERXJMI-UHFFFAOYSA-N

Application

The presence of electrolyte additives enhances the performance of lithium ion batteries by improving their thermal stability and ionic conductivity. Additives also reduce the flammability of electrolytes and provide overcharge protection. 2-Propynyl methanesulfonate finds application as additive in lithium ion batteries.

Preparation Note

  • These additives have low water content (less than 100 ppm), please handle under inert and moisture free environment (glove box).
  • Keep containers tightly closed.
  • Keep away from heat and ignition sources.
  • Store in a cool and dry place.
  • Avoid storing together with oxidizers.

Legal Information

Product of MU Ionic Solutions Corp

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hogstrom K, et al.
The Journal of Physical Chemistry C, 118, 12649-12649 (2014)
Zhang S.
Journal of Power Sources, 162, 1379-1379 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service