Skip to Content
Merck
All Photos(3)

Key Documents

402885

Sigma-Aldrich

p-Toluenesulfonic acid monohydrate

ACS reagent, ≥98.5%

Synonym(s):

4-Methylbenzenesulfonic acid monohydrate, 4-Toluenesulfonic acid monohydrate, PTSA monohydrate, TsOH monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3H · H2O
CAS Number:
Molecular Weight:
190.22
Beilstein:
3568023
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

5.9 (vs air)

Assay

≥98.5%

form

solid

clarity of soln

passes test

impurities

9.5-11.5% water

ign. residue

≤0.1%

mp

103-106 °C (lit.)

solubility

H2O: 10g + 50 mL, clear, colorless

anion traces

sulfate (SO42-): ≤0.3%

cation traces

Fe: ≤0.01%
Na: ≤0.002%
heavy metals (as Pb): ≤0.001%

functional group

tosylate

SMILES string

[H]O[H].Cc1ccc(cc1)S(O)(=O)=O

InChI

1S/C7H8O3S.H2O/c1-6-2-4-7(5-3-6)11(8,9)10;/h2-5H,1H3,(H,8,9,10);1H2

InChI key

KJIFKLIQANRMOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

p-Toluenesulfonic acid monohydrate, an oxonium salt, is an inexpensive and easy to handle organic catalyst used in organic synthesis. The study of its crystalline structure shows that it is monoclinic with P21/c space group. Its solubility in aqueous sulfuric acid solutions has been studied.

Application

p-Toluenesulfonic acid monohydrate (p-TsOH·H2O) may be used as a catalyst in the synthesis of the following:
  • Unsymmetrical benzils.
  • Highly substituted piperidines.
  • 1,3,5-Trisubstituted benzenes by trimerization of alkynes.
  • Triazoloquinazolinone and benzimidazoquinazolinone derivatives.
  • 1,3,5-Trisubstituted pyrazoles derivatives.
  • Selenated ketene dithioacetals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solubilities of p-Toluenesulfonic Acid Monohydrate and Sodium p-Toluenesulfonate in Aqueous Sulfuric Acid Solutions and Its Application for Preparing Sodium p-Toluenesulfonate.
Zhao W, et al.
Industrial & Engineering Chemistry Research, 52(51), 18466-18471 (2013)
One-pot multicomponent synthesis of highly substituted piperidines using p-toluenesulfonic acid monohydrate as catalyst.
Sajadikhah SS, et al.
Monatshefte fur Chemie / Chemical Monthly, 143(6), 939-945 (2012)
Catalytic systems containing p-toluenesulfonic acid monohydrate catalyzed the synthesis of triazoloquinazolinone and benzimidazoquinazolinone derivatives.
Mousavi MR and Maghsoodlou MT.
Monatshefte fur Chemie / Chemical Monthly, 145(12), 1967-1973 (2014)
Br?nsted acid-catalyzed phenylselenenylation of internal olefins.
Wu P, et al.
Tetrahedron Letters, 56(19), 2488-2491 (2015)
An efficient approach to the cyclotrimerisation of alkynes: solvent-free synthesis of 1, 3, 5-trisubstituted benzenes using p-toluenesulfonic acid monohydrate.
Gao Q, et al.
ARKIVOC (Gainesville, FL, United States), 3, 49-60 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service