Skip to Content
Merck
All Photos(2)

Documents

718203

Sigma-Aldrich

Allyl methyl sulfone

greener alternative

96%

Synonym(s):

3-(Methylsulfonyl)-1-propene, MAS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2S
CAS Number:
Molecular Weight:
120.17
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.472

bp

244 °C (lit.)

density

1.171 g/mL at 25 °C

application(s)

battery manufacturing

greener alternative category

SMILES string

CS(=O)(=O)CC=C

InChI

1S/C4H8O2S/c1-3-4-7(2,5)6/h3H,1,4H2,2H3

InChI key

WOPDMJYIAAXDMN-UHFFFAOYSA-N

General description

Allyl methyl sulfone is an organosulfur compound. It can be used as a solvent or an additive for electrochemical applications.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of metal oxide gas sensor in electronic nose to monitor odor profiles of garlic scape
Messina V, et al.
IEEE Sensors Journal, 14(6), 1765-1769 (2014)
Attempt to detect garlic allyl sulphides from saliva after consumption of garlic tablets using GC-MS
Watson CJ, et al.
Journal of Microbial & Biochemical Technology, 5, 081-083 (2013)
Taichi Onishi et al.
Scientific reports, 8(1), 1692-1692 (2018-01-28)
Disc1 is a susceptibility gene for psychiatric disorders including schizophrenia. It has been suggested that excess transmission through dopamine type 2 receptors (D2Rs) in the striatum is an underlying mechanism of pathogenesis. In this study, we used super-resolution microscopy to
Sveta Zhiraslanovna Ozkan et al.
Polymers, 11(7) (2019-07-25)
Hybrid nanocomposites based on electroactive polydiphenylamine-2-carboxylic acid (PDPAC) and single-walled carbon nanotubes (SWCNTs) were obtained for the first time. Polymer-carbon nanomaterials were synthesized via in situ oxidative polymerization of diphenylamine-2-carboxylic acid (DPAC) in the presence of SWCNTs by two different
Youngteak Yoon et al.
Journal of neurogenetics, 33(3), 179-189 (2019-06-08)
The way in which the central nervous system (CNS) governs animal movement is complex and difficult to solve solely by the analyses of muscle movement patterns. We tackle this problem by observing the activity of a large population of neurons

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service