Skip to Content
Merck
All Photos(9)

Documents

746428

Sigma-Aldrich

Potassium iodide

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
anhydrous

Quality Level

vapor pressure

1 mmHg ( 745 °C)

product line

Redi-Dri

Assay

≥99%

form

crystals

quality

free-flowing

impurities

≤0.001% N compounds
≤0.005% insolubles

loss

≤0.2% loss on drying

pH

6.0-9.2 (25 °C, 5%)

mp

681 °C (lit.)

anion traces

chloride, bromide (as Cl)-: ≤0.01%
iodate (IO3-): ≤3 ppm
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤3 ppm
Mg: ≤0.001%
Na: ≤0.005%
heavy metals (as Pb): ≤5 ppm

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium iodide is a water soluble inorganic salt that can be prepared by reacting hydrogen iodide and potassium bicarbonate.

Redi-Dri technology eliminates clumping and the need for complicated procedures to keep your salts free-flowing. Keeping your salts dry helps improve your results by giving accurate measurements and easier-to-handle salts and buffers. Rather than drying your salts in an oven or desiccator or breaking them apart manually, Redi-Dri ensures that your salts will remain easy-to-use for months without worrying about anti-caking agents. Even after repeated openings you can be assured that your results are always accurate. Redi-Dri saves you time, energy and money by keeping your salts usable for longer!

Application

Potassium iodide may be used as a catalyst in the following processes:
  • Conversion of aromatic primary amines to the corresponding nitro compounds in the presence of tert-butyl hydroperoxide as oxidant.
  • Conversion of aryl halide to aryl iodides in the presence of nickel catalysts.
  • Preparation of sulfonated oxindoles from activated alkenes and sulfonylhydrazides in water and in the presence of 8-crown-6 and tert-butyl hydroperoxide.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing Chu et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 7(8), 1903592-1903592 (2020-04-25)
Congenital heart disease (CHD) is the major cause of morbidity/mortality in infancy and childhood. Using a mouse model to uncover the mechanism of CHD is essential to understand its pathogenesis. However, conventional 2D phenotyping methods cannot comprehensively exhibit and accurately
J B Sterling et al.
Journal of the American Academy of Dermatology, 43(4), 691-697 (2000-09-27)
Potassium iodide (KI) is a useful drug in the dermatologic armamentarium. It is successfully used for inflammatory dermatoses, most notably erythema nodosum, subacute nodular migratory panniculitis, nodular vasculitis, erythema multiforme, and Sweet's syndrome. KI is also successfully used for cutaneous
Pietro Santulli et al.
Human reproduction (Oxford, England), 30(1), 49-60 (2014-11-08)
Are protein oxidative stress markers [thiols, advanced oxidation protein products (AOPP), protein carbonyls and nitrates/nitrites] in perioperative peritoneal fluid higher in women with histologically proven endometriosis when compared with endometriosis-free controls? Protein oxidative stress markers are significantly increased in peritoneal
Ilzira A Minigalieva et al.
International journal of molecular sciences, 16(9), 22555-22583 (2015-09-24)
Stable suspensions of NiO and Mn₃O₄ nanoparticles (NPs) with a mean (±s.d.) diameter of 16.7±8.2 and 18.4±5.4 nm, respectively, purposefully prepared by laser ablation of 99.99% pure nickel or manganese in de-ionized water, were repeatedly injected intraperitoneally (IP) to rats
Christoph Reiners et al.
Radiation and environmental biophysics, 52(2), 189-193 (2013-03-12)
Thyroid cancer in children and adolescents has to be considered as the most severe health consequence of a nuclear reactor emergency with release of radioiodine into the atmosphere. High doses of potassium iodide are effective to block radioiodine thyroid uptake

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service