Skip to Content
Merck
All Photos(1)

Documents

764582

Sigma-Aldrich

Polyethylenimine, linear

average Mn 5,000, PDI ≤1.3

Synonym(s):

PEI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2NH)n
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 5,000

mp

54-59 °C

PDI

≤1.3

storage temp.

2-8°C

InChI

1S/C2H5N/c1-2-3-1/h3H,1-2H2

InChI key

NOWKCMXCCJGMRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

All polyethylene imine polymers are hydrophilic and may contain approx. 30% hydrated water.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Hoffmann et al.
International journal of molecular sciences, 21(6) (2020-03-28)
Highly efficient, biocompatible, and fast nucleic acid delivery methods are essential for biomedical applications and research. At present, two main strategies are used to this end. In non-viral transfection liposome- or polymer-based formulations are used to transfer cargo into cells
Prashanth K B Nagesh et al.
Cancers, 10(9) (2018-08-29)
The therapeutic application of microRNA(s) in the field of cancer has generated significant attention in research. Previous studies have shown that miR-205 negatively regulates prostate cancer cell proliferation, metastasis, and drug resistance. However, the delivery of miR-205 is an unmet
Laura R H Ip et al.
Oncotarget, 6(12), 10548-10562 (2015-04-15)
Treatment options for ovarian cancer patients remain limited and overall survival is less than 50% despite recent clinical advances. The lipid phosphatase inositol polyphosphate 4-phosphatase type II (INPP4B) has been described as a tumor suppressor in the PI3K/Akt pathway with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service