Merck
  • Loss of INPP4B causes a DNA repair defect through loss of BRCA1, ATM and ATR and can be targeted with PARP inhibitor treatment.
Loss of INPP4B causes a DNA repair defect through loss of BRCA1, ATM and ATR and can be targeted with PARP inhibitor treatment.
Oncotarget (2015-04-15)
Laura R H Ip, George Poulogiannis, Felipe Cia Viciano, Junko Sasaki, Satoshi Kofuji, Victoria J Spanswick, Daniel Hochhauser, John A Hartley, Takehiko Sasaki, Christina A Gewinner
ABSTRACT
Treatment options for ovarian cancer patients remain limited and overall survival is less than 50% despite recent clinical advances. The lipid phosphatase inositol polyphosphate 4-phosphatase type II (INPP4B) has been described as a tumor suppressor in the PI3K/Akt pathway with loss of expression found most pronounced in breast, ovarian cancer and melanoma. Using microarray technology we identified a DNA repair defect in INPP4B-deficient cells, which we further characterized by comet assays and quantification of γH2AX, RAD51 and 53BP1 foci formation. INPP4B loss resulted in significantly increased sensitivity towards PARP inhibition, comparable to loss of BRCA1 in two- and three-dimensional in vitro models, as well as in in vivo xenograft models. Mechanistically, we discovered that INPP4B forms a protein complex with the key players of DNA repair, ATR and BRCA1, in GST pulldown and 293T overexpression assays, and INPP4B loss affects BRCA1, ATM and ATR protein stability resulting in the observed DNA repair defect. Given that INPP4B loss has been found in 40% of ovarian cancer patients, this study provides the rationale for establishing INPP4B as a biomarker of PARP inhibitor response, and consequently offers novel therapeutic options for a significant subset of patients. Loss of the tumor suppressor inositol polyphosphate 4-phosphatase type II (INPP4B) results in a DNA repair defect due to concomitant loss of BRCA1, ATR and ATM and can be therapeutically targeted with PARP inhibitors.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Poly(ethyleneimine) solution, average Mw 1800, 50 wt. % in H2O
Sigma-Aldrich
Poly(ethyleneimine) solution, average Mn ~1,200, average Mw ~1300 by LS, 50 wt. % in H2O
Sigma-Aldrich
Poly(ethyleneimine) solution, average Mn ~60,000 by GPC, average Mw ~750,000 by LS, 50 wt. % in H2O
Sigma-Aldrich
Polyethylenimine, linear, average Mn 10,000, PDI ≤1.3
Sigma-Aldrich
Polyethylenimine, linear, average Mn 5,000, PDI ≤1.3
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Polyethylenimine, linear, average Mn 2100, PDI <1.3
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Anti-ATM Antibody, Upstate®, from rabbit