Skip to Content
Merck
All Photos(1)

Documents

764604

Sigma-Aldrich

Polyethylenimine, linear

average Mn 2100, PDI <1.3

Synonym(s):

PEI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2NH)n
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

mol wt

average Mn 2100

mp

59-64 °C

transition temp

Tm 59-64 °C

PDI

<1.3

storage temp.

2-8°C

InChI

1S/C2H5N/c1-2-3-1/h3H,1-2H2

InChI key

NOWKCMXCCJGMRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyethylenimine(PEI) is a synthetic cationic polymerwith high charge density and chain flexibility. It is the most widelyinvestigated cationic polymer for non-viral delivery of nucleic acids. LinearPEI can be an effective carrier for a wide range of gene medicine including DNAplasmids, small interfering RNAs, and mRNAs. It protects the DNA from theacidic environment and ensures the release of DNA to the cytoplasm.

Application

  • Water-soluble linear poly (ethylenimine) as a superior bifunctional binder for lithium–sulfur batteries: Demonstrates the utility of water-soluble linear polyethylenimine as an effective bifunctional binder that enhances the cycle stability and efficiency of lithium-sulfur batteries, pivotal for advancing battery technology (Liao et al., 2018).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Feng et al.
Engineering in life sciences, 20(8), 320-330 (2020-08-11)
As the main immunogen that could stimulate neutralized antibody in pigs, recombinant E2 protein of CSFV was expressed in CHO-dhfr-cells driven by endogenous Txnip promoter from Chinese hamster. Different fragments of Txnip promoter were amplified by PCR from isolated genomic
Laura R H Ip et al.
Oncotarget, 6(12), 10548-10562 (2015-04-15)
Treatment options for ovarian cancer patients remain limited and overall survival is less than 50% despite recent clinical advances. The lipid phosphatase inositol polyphosphate 4-phosphatase type II (INPP4B) has been described as a tumor suppressor in the PI3K/Akt pathway with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service