Skip to Content
Merck
All Photos(1)

Key Documents

408700

Sigma-Aldrich

Poly(ethyleneimine) solution

average Mw 1800, 50 wt. % in H2O

Synonym(s):

Ethyleneimine polymer solution, PEI

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

mol wt

average Mw 1800

concentration

50 wt. % in H2O

refractive index

n20/D 1.453

density

1.08 g/mL at 25 °C

SMILES string

N1CC1.NCCN

InChI

1S/C2H8N2.C2H5N/c3-1-2-4;1-2-3-1/h1-4H2;3H,1-2H2

InChI key

SFLOAOINZSFFAE-UHFFFAOYSA-N

General description

Poly(ethyleneimine) (PEI) is a positivelycharged polymeric amine with a linear or branched structure. It has chelating properties with a variety of metal ions. It is highlysoluble in water and behaves as an effective base in aqueous solutions. Due toits superior physicochemical properties, it is widely used in the field ofbiomedicine, water purification, bioimaging, gene delivery, and self-healingmaterials.

Application

Poly(ethyleneimine) solution can be used for surface modification of cation exchange resins. The modified resins exhibited improved selectivity towards transition elements. Due to repulsion effects within the cationic layer, the selectivity for alkaline ions decreases. As a result, this resin can be applied to purify industrial waste by column sorption.

It can be used as a precursor to prepare polyelectrolyte multilayers for the promotion of cell adhesion and growth on implant surfaces.

PEI can act as a reactive template and stabilizing agent in the preparation of metal nanoparticles in aqueous media. The amino groups in the PEI can coordinate with metal ions dissolved in water and reduce their aggregation capability. This results in the formation of smaller nanoparticles.

Features and Benefits

  • It has repetitive structural units and aflexible polymer chain.
  • Due to the accessibility of a huge number ofamino groups on PEI, it can be modified or functionalized for specific featuressuch as sensitivity, specificity, and biocompatibility.
  • Highwater solubility.

Physical form

Branched polymer

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathon M Muncie et al.
Developmental cell, 55(6), 679-694 (2020-11-19)
Embryogenesis is directed by morphogens that induce differentiation within a defined tissue geometry. Tissue organization is mediated by cell-cell and cell-extracellular matrix (ECM) adhesions and is modulated by cell tension and tissue-level forces. Whether cell tension regulates development by modifying
Laura R H Ip et al.
Oncotarget, 6(12), 10548-10562 (2015-04-15)
Treatment options for ovarian cancer patients remain limited and overall survival is less than 50% despite recent clinical advances. The lipid phosphatase inositol polyphosphate 4-phosphatase type II (INPP4B) has been described as a tumor suppressor in the PI3K/Akt pathway with
Diana F Silva et al.
Journal of neuroinflammation, 17(1), 332-332 (2020-11-07)
After decades of research recognizing it as a complex multifactorial disorder, sporadic Alzheimer's disease (sAD) still has no known etiology. Adding to the myriad of different pathways involved, bacterial neurotoxins are assuming greater importance in the etiology and/or progression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service