Skip to Content
Merck
All Photos(3)

Key Documents

448869

Sigma-Aldrich

Chitosan

low molecular weight

Synonym(s):

Deacetylated chitin, Poly(D-glucosamine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.23
Pricing and availability is not currently available.

form

powder

mol wt

50,000-190,000 Da (based on viscosity)

viscosity

20-300 cP, 1 wt. % in 1% acetic acid(25 °C, Brookfield)(lit.)

solubility

dilute aqueous acid: soluble

SMILES string

N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O)O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4N)O)O[C@@H]5O[C@@H]([C@H]([C@@H]([C@H]5N)O)O[C@@H]6O[C@@H]([C@H]([C@@H]([C@H]6N)O)O[C@@H]7O[C@@H]([C@H]([C@@H]([C@H]7N)O)O[C@@H]8O[C@@H]([C@H]([C@@H]([C@H]8N)O)O[C@@H]9O[C@@H]([C@H](

InChI

1S/C56H103N9O39/c1-87-56(86)65-28-38(84)46(19(10-74)96-55(28)104-45-18(9-73)95-49(27(64)37(45)83)97-39-12(3-67)88-47(85)20(57)31(39)77)103-54-26(63)36(82)44(17(8-72)94-54)102-53-25(62)35(81)43(16(7-71)93-53)101-52-24(61)34(80)42(15(6-70)92-52)100-51-23(60)33(79)41(14(5-69)91-51)99-50-22(59)32(78)40(13(4-68)90-50)98-48-21(58)30(76)29(75)11(2-66)89-48/h11-55,66-85H,2-10,57-64H2,1H3,(H,65,86)/t11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48+,49+,50+,51+,52+,53+,54+,55+/m1/s1

InChI key

FLASNYPZGWUPSU-SICDJOISSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chitosan may be used as a flocculant, in protein precipitation, encapsulating agent and aqueous thickener.
Forms gels with multivalent anions. Gives clear solutions that dry to strong, clear films.

Features and Benefits

Biocompatible, antibacterial and environmentally friendly polyelectrolyte with a variety of applications including water treatment, chromatography, additives for cosmetics, textile treatment for antimicrobial activity, novel fibers for textiles, photographic papers, biodegradable films, biomedical devices, and microcapsule implants for controlled release in drug delivery.
Biocompatible, antibacterial and environmentally friendly polyelectrolyte.

Physical form

75-85% deacetylated

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-Hui Yang et al.
BioMed research international, 2014, 789591-789591 (2014-04-24)
Chitosan nanoparticle, a biocompatible material, was used as a potential drug delivery system widely. Our current investigation studies were the bioeffects of the chitosan nanoparticle uptake by liver cells. In this experiment, the characterizations of chitosan nanoparticles were measured by
Tsung-Lin Yang et al.
Journal of cellular and molecular medicine, 13(9A), 2853-2863 (2008-07-17)
Chitosan is a widely used biocompatible biomaterial in the tissue regeneration, but its utility and application in the tissue morphogenesis of salivary gland remains unclear. The study aimed to explore the effects of chitosan on the epithelial morphogenesis of submandibular
Ajay Vijayakumar et al.
AAPS PharmSciTech, 18(3), 875-883 (2016-07-03)
The objective of this study was to formulate and characterize properties of solid lipid nanoparticle (SLN) dispersion containing quercetin. SLN was prepared by ultrasonication method using tripalmitin and lecithin as lipid core and then the surface was coated with chitosan.
Wei-Hsuan Lo et al.
Molecules (Basel, Switzerland), 25(21) (2020-11-07)
(1) Background: Few antifungal drugs are currently available, and drug-resistant strains have rapidly emerged. Thus, the aim of this study is to evaluate the effectiveness of the antifungal activity from a combinational treatment of chitosan with a clinical antifungal drug
Jimena S Gonzalez et al.
Polymers, 11(4) (2019-04-20)
Self-assembly of natural polymers constitute a powerful route for the development of functional materials. In particular, layer-by-layer (LBL) assembly constitutes a versatile technique for the nanostructuration of biobased polymers into multilayer films. Gelatin has gained much attention for its abundance

Questions

1–10 of 20 Questions  
  1. Dear Sir, could I make a dilution serious of HPLC Chitosan and analysis through HPLC for quantification and calibration curve? and what molecular weight should I use for 250 microgram/mL stock solution?

    1 answer
    1. Yes, a dilution series of HPLC-grade chitosan can be made for HPLC analysis to establish a calibration curve for quantification. However, no molecular weight information is available.

      Helpful?

  2. Dear sir Could we get the (HNMR) and viscosity testing for the determination of the degree of deacetylated chitosan as we used the one with "(50,000–190,000 Da and degree of deacetylation equal or greater than 70%.Polymers Journal requested it.

    1 answer
    1. The result for the degree of deacetylation is provided on the certificate of analysis for each lot of this product. Unfortunately, the assay method may not be shared.

      Helpful?

  3. Can i use this Chitosan for fruit coating?

    1 answer
    1. This product is intended for research use only and is not food grade. Therefore it is not recommended for a fruit coating application.

      Helpful?

  4. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  5. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  6. We need to confirm if this Chitosan (Catalog #448869) is of animal origin (shellfish) or synthetic.

    1 answer
    1. The source information is located on the Certificate of Origin and may vary from lot to lot. Recent lot shows the source of this product is from the shells of Pandalus Borealis, Cold Water Shrimp.

      Helpful?

  7. What is the molecular weight (Mw) of products 448869, 448877, or 419419?

    1 answer
    1. The molecular weight (Mw) of products 448869, 448877, and 419419 are approximately 150-250 kDa, 250-300 kDa, and 340-360 kDa, respectively. These values are calculated based on the viscosity values provided on the product data sheets and the corresponding table of viscosity vs Mw.

      Helpful?

  8. What are the Mark-Houwink–Sakurada parameters (K and alpha) you use for determination of molecular weight through viscosity? Thank you!

    1 answer
    1. The Certificates of Analysis(CofA) for each lot of Product 448869, a low molecular weight Chitosan, do not report a molecular weight. However, the viscosity in 1 percent acetic acid (abbreviated HAc or HOAc) is on the CofA. Values for K and alpha for high molecular weight Chitosan have been published, which might also apply to a low molecular weight Chitosan; see Calculation of Mark–Houwink–Sakurada (MHS) equation viscometric constants for chitosan in any solvent–temperature system using experimentally reported viscometric constants data, by Mohammed R. Kasaai. Carbohydrate Polymers, 68(3), 477-488 (2007).

      Helpful?

  9. Is this analytical grade?

    1 answer
    1. This product is not considered analytical grade. It is intended for general research use. Currently, no analytical grade product option is available.

      Helpful?

  10. Is it research or food grade? Thanks

    1 answer
    1. This product is intended for research use only. Currently, there is no food-grade product option available

      Helpful?

1–10 of 20 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service