Skip to Content
Merck
All Photos(2)

Key Documents

757365

Sigma-Aldrich

Lithium nickel dioxide

powder, <3 μm particle size (BET), ≥98% trace metals basis

Synonym(s):

LNO, Lithium nickel oxide, Lithium nickelate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNiO2
CAS Number:
Molecular Weight:
97.63
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

Quality Level

Assay

≥98% trace metals basis

form

powder

mol wt

Mw 97.63 g/mol

composition

LiNiO2

particle size

<3 μm (BET)

mp

>1,000 °C (lit.)

density

4.62 g/cm3 (lit.)

application(s)

battery manufacturing

SMILES string

[Ni](=O)[O-].[Li+]

InChI

1S/Li.Ni.2O/q+1;;;-1

InChI key

VROAXDSNYPAOBJ-UHFFFAOYSA-N

Related Categories

General description

Lithium nickel dioxide (LNO) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.

Application

Cathode material with high capacity and good reversibility for rechargeable Lithium ion batteries. Sub-micron particle size results in increased surface area of electrodes, thus improving their performance.
LNO can be used as a cathode material with a high discharge voltage of 4V and a high diffusion coefficient of ~10−8-10−10 cm2s−1. It has a high specific capacity and can be used the fabrication of lithium-ion batteries.

Legal Information

Product of Engi-Mat Co.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A Inhalation - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical characterization of a lithiated mixed nickel-cobalt oxide (LiNi 0.5 Co 0.5 O 2) prepared by sol-gel process
Croce F, et al.
Ionics, 3(5-6), 390-395 (1997)
Thackeray, M. M.; et al.
Journal of Materials Chemistry, 17, 3112-3112 (2007)
Synthesis and electrode performance of layered nickel dioxide containing alkaline ions
Arai H, et al.
Electrochimica Acta, 50(9), 1821-1828 (2005)
Broussely, M.; et al.
Journal of Power Sources, 54, 109-109 (1995)
Towards greener and more sustainable batteries for electrical energy storage
Larcher D and Tarascon J
Nature Chemistry, 7(1), 19-19 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service