Skip to Content
Merck
All Photos(2)

Key Documents

254290

Sigma-Aldrich

Lithium niobate

99.9% trace metals basis

Synonym(s):

Lithium metaniobatem, Lithium niobium oxide, Lithium niobium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNbO3
CAS Number:
Molecular Weight:
147.85
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

core: niobium
reagent type: catalyst

particle size

200 mesh

SMILES string

[Li+].[O-][Nb](=O)=O

InChI

1S/Li.Nb.3O/q+1;;;;-1

InChI key

GQYHUHYESMUTHG-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hector de Castilla et al.
Materials (Basel, Switzerland), 12(16) (2019-08-10)
Piezoelectric materials have the intrinsic reversible ability to convert a mechanical strain into an electric field and their applications touch our daily lives. However, the complex physical mechanisms linking mechanical and electrical properties make these materials hard to understand. Computationally
Tao Chen et al.
Applied optics, 51(28), 6881-6885 (2012-10-04)
We experimentally demonstrated a compact fiber laser-pumped multichannel PPMgLN-based optical parametric oscillator (OPO) generating total OPO output power of 15.8, 15.2, 14.2, 12.9, and 8.8 W with idler output power of 4.7, 4.3, 4.1, 3.3, and 2.1 W at the
W Horn et al.
Optics express, 20(24), 26922-26928 (2012-11-29)
We report the fabrication of femtosecond laser-induced, first-order waveguide Bragg gratings in lithium niobate in the low repetition rate regime. Type-II waveguides are written into an x-cut lithium niobate wafer and structured periodically to achieve narrowband reflections at wavelengths around
Swapnesh Panigrahi et al.
Nature communications, 11(1), 549-549 (2020-01-30)
High-frequency demodulation of wide area optical signals in a snapshot manner remains a technological challenge. If solved, it could open tremendous perspectives in 3D imaging, vibrometry, free-space communications, automated vision, or ballistic photon imaging in scattering media with numerous applications
Jesper B Christensen et al.
Molecules (Basel, Switzerland), 26(3) (2021-01-29)
We report on the use of quartz-enhanced photoacoustic spectroscopy for continuous carbon-dioxide measurements in humid air over a period of six days. The presence of water molecules alters the relaxation rate of the target molecules and thus the amplitude of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service