Skip to Content
Merck
All Photos(3)

Key Documents

481793

Sigma-Aldrich

Nickel(II) oxide

greener alternative

≥99.995% trace metals basis

Synonym(s):

Mononickel oxide, Nickel Oxide Sinter 75, Nickel oxide, Nickelous oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiO
CAS Number:
Molecular Weight:
74.69
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.995% trace metals basis

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

dark green

density

6.67 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Ni]=O

InChI

1S/Ni.O

InChI key

GNRSAWUEBMWBQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for Molecular Solar Thermal Energy Storage Systems (MOST). Click here for more information.

Application


  • Incorporation of Nanocatalysts for the Production of Bio-Oil from Staphylea holocarpa Wood.: This research explores the application of nanocatalysts, including Nickel(II) oxide, in the production of bio-oil, showcasing its role in enhancing the energy yield and efficiency of the bio-oil production process (Li et al., 2022).


Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 1A Inhalation - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genqiang Zhang et al.
Nanoscale, 5(3), 877-881 (2012-12-15)
One-dimensional hierarchical hollow nanostructures composed of NiO nanosheets are successfully synthesized through a facile carbon nanofiber directed solution method followed by a simple thermal annealing treatment. With the advantages of high electro-active surface area, carbon nanofiber supported robust structure and
N Sattarahmady et al.
Biosensors & bioelectronics, 48, 197-202 (2013-05-21)
Lichen-like nickel oxide nanostructure was synthesized by a simple method and characterized. The nanostructure was then applied to modify a carbon paste electrode and for the fabrication of a sensor, and the electrocatalytic oxidation of acetylcholine (ACh) on the modified
Daniele Perego et al.
Nanotechnology, 24(4), 045302-045302 (2013-01-08)
In the present paper, a novel method to fabricate ordered arrays of Au/NiO/Au nanowires is described, with the aim of filling the gap between the fundamental study of the electrical properties of scattered single nanowires and the engineered fabrication of
Liqiang Luo et al.
Colloids and surfaces. B, Biointerfaces, 102, 307-311 (2012-09-26)
A highly sensitive and selective nonenzymatic glucose sensor based on electrodepositing NiO nanoparticles on ordered mesoporous carbon (named as NiO/OMC) modified glassy carbon electrode (GCE) was constructed. The synthesized OMC was characterized by X-ray diffraction, and the morphology images of
Huaixiang Li et al.
Biosensors & bioelectronics, 47, 225-230 (2013-04-16)
A novel photoelectrochemical hydrogen peroxide (H2O2) sensor was constructed with platinum (Pt) and nickel hydroxyl-oxide (NiOOH) double layers modified n-silicon electrode (NiOOH/Pt/n-n(+)-Si). About 40nm Pt layer and about 100nm Ni layer were successively coated on the front surface of n-n(+)-Si

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service