Skip to Content
Merck
All Photos(1)

Key Documents

72262

Sigma-Aldrich

Nickel (IV) oxide

technical, oxidizing agent, ~30% active peroxide basis

Synonym(s):

Nickel dioxide, Nickel oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NiO2
CAS Number:
Molecular Weight:
90.69
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

grade

oxidizing agent
technical

Quality Level

form

powder

reaction suitability

core: nickel
reagent type: catalyst
reagent type: oxidant

concentration

~30% (active peroxide)

SMILES string

O=[Ni]O[Ni]=O

InChI

1S/2Ni.3O

InChI key

PZFKDUMHDHEBLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • One-dimensional oxide-metal hybrid structures and site-specific enhanced reactivity for CO oxidation
  • Microwave induced catalytic degradation of crystal violet

Used to study differences in reactivity trends for carbon monoxide catalyzed oxidation caused by anionic transition metal oxide clusters

Reagent:
  • For solvent-free oxidation of alcohols†
  • For chemoselective oxidation of organic compounds using inductive heating
  • To improve the synthesis of methoxy cephalosporin′s intermediate

Other Notes

Oxidant for the transformation of alcohols into acids and hydrazones into diazoalkanes

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 1A Inhalation - Ox. Sol. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Nakagawa et al.
The Journal of Organic Chemistry, 27, 1597-1597 (1962)
Rut Sanchis et al.
Materials (Basel, Switzerland), 12(18) (2019-09-12)
Different nickel catalysts have been tested for the transformation of levulinic acid into γ-valerolactone using an easy hydrothermal method, taking advantage of the properties of the high temperature water. A metallic nickel catalyst derived from NiO synthesized by a nanocasting
Chien-Chen Diao et al.
Nanomaterials (Basel, Switzerland), 10(4) (2020-04-03)
In this study, a p-type 2 at% lithium-doped nickel oxide (abbreviation L2NiO) solution was prepared using Ni(NO3)2·6H2O, and LiNO3·L2NiO thin films were deposited using an atomizer by spraying the L2NiO solution onto a glass substrate. The sprayed specimen was heated
Saiganesh Srihasam et al.
Biomolecules, 10(1) (2020-01-16)
In the present study, economically viable NiO nanoparticles were produced by biogenic preparation using stevia leaf broth and their in-vitro antioxidant and antimicrobial activities were evaluated. The properties of the prepared NiO nanoparticles were confirmed by analytical techniques such as
Zachary Daniels et al.
Materials (Basel, Switzerland), 12(18) (2019-09-11)
Described herein is a novel method, Reduction Expansion Synthesis-Sintered Metal (RES-SM), to create a sintered metal body of a designed shape at ambient pressure, hundreds of degrees below the metal melting temperature. The precursor to the metal part is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service