Skip to Content
Merck
All Photos(4)

Key Documents

637130

Sigma-Aldrich

Nickel(II) oxide

greener alternative

nanopowder, <50 nm particle size (TEM), 99.8% trace metals basis

Synonym(s):

Nickel monooxide, Nickelous oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiO
CAS Number:
Molecular Weight:
74.69
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.8% trace metals basis

form

nanopowder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

green to black

particle size

<50 nm (TEM)

density

6.67 g/mL at 25 °C (lit.)

bulk density

0.51 g/mL

application(s)

battery manufacturing

greener alternative category

SMILES string

[Ni]=O

InChI

1S/Ni.O

InChI key

GNRSAWUEBMWBQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) oxide (NiO) is a metal oxide based nanomaterial with a good semiconducting property. Nanosized nickel oxide can be found in a variety of morphologies which include nanoflowers, spheres, wires, and tubes. It exhibits high performance in applications which require charge transfer and charge transport based processes. It can be prepared by a variety of physical and thermal methods such as sol-gel, hydrothermal and solvothermal techniques.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for Molecular Solar Thermal Energy Storage Systems (MOST). Click here for more information.

Application

NiO can be used for a variety for energy and coating based applications such as lithium ion batteries, solar cells, electrochromic coatings, semiconducting films, and gas based sensors.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Carc. 1A Inhalation - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hierarchical NiO hollow microspheres assembled from nanosheet-stacked nanoparticles and their application in a gas sensor
Zhu G, et al.
Royal Society of Chemistry Advances, 2(10), 4236-4241 (2012)
PbS-NiO nanocomposite material with enhanced magnetic, photocatalytic and antifungal properties
Suganya M, et al.
Materials Science and Engineering, B, 229(1), 118-125 (2018)
Fabrication of NiO nanowall electrodes for high performance lithium ion battery
Varghese B, et al.
Chemistry of Materials, 20(10), 3360-3367 (2008)
A p-type NiO-based dye-sensitized solar cell with an open-circuit voltage of 0.35 V
Gibson EA, et al.
Angewandte Chemie (International ed. in English), 48(24), 4402-4405 (2009)
A novel precursor in preparation and characterization of nickel oxide nanoparticles via thermal decomposition approach
Salavati-Niasari M, et al.
Journal of alloys and compounds, 493(1-2), 163-168 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service