715468
Barium tert-butoxide
85%
Synonym(s):
Barium di-t-butoxide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
85%
form
powder
mp
>200 °C
storage temp.
2-8°C
SMILES string
CC(C)(C)O[Ba]OC(C)(C)C
InChI
1S/2C4H9O.Ba/c2*1-4(2,3)5;/h2*1-3H3;/q2*-1;+2
InChI key
SLPLCLDJTNLWPW-UHFFFAOYSA-N
General description
Barium tert-butoxide is a Brønsted base catalyst that can be used as a precursor of various barium catalysts like barium aryloxide. It can also be used in the preparation of barium diphenylmethanide, applicable in the organometallic chemistry.
Application
Barium tert-butoxide can be used:
- As a catalyst along with various phenolic ligands in the aldol reactions of amides with aldehydes and ketones.
- To prepare yttrium and copper metal complexes by reacting with Y(thd)3 and Cu(thd)2, where “thd” is 2,2,6,6-tetramethylheptane-3,5-dione.
- As an initiator along with an alkyl lithium in the butadiene copolymerization reaction.
- In the iridium catalyzed allylic alkylation of non-stabilized enolates.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 2 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Reactivity of mixed-metal BaCu and BaY β-diketonatoalkoxides with lewis bases: Molecular structures of [Ba2Cu (μ3, η2-OCHMeCH2NMe2) 2 (μ, η2-thd) 2 (η2-thd) 2 (PriOH2],[Cu3 (μ3-OBut) 2 (μ-OBut)(η2-thd) 3] and [Ba (η2-thd) 2 (TMEDA) 2]
Polyhedron, 15(4), 577-589 (1996)
Highly anti-Selective Catalytic Aldol Reactions of Amides with Aldehydes
Journal of the American Chemical Society, 128(27), 8704-8705 (2006)
Iridium-catalyzed diastereo-, enantio-, and regioselective allylic alkylation with prochiral enolates
ACS Catalysis, 6(9), 6207-6213 (2016)
Barium tert-butoxide
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2009)
Accessible microstructures of polybutadiene by anionic polymerization
Polymer, 153(4), 103-122 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service