Skip to Content
Merck
All Photos(5)

Key Documents

F9378

Sigma-Aldrich

Ficoll® 400

Synonym(s):

Polysucrose 400

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

Application

Ficoll® 400 is used together with sodium diatrizoate (Paque) to create Ficoll-Paque, a density mixture, used for the separation of cells and organelles, especially blood derived lymphocytes and monocytes. Ficoll 400 may be used for the development of new cell and organelle separation systems. Ficoll 400 is used in tissue engineering and macromolecular conformation research for the development, evaluation and use of macromolecular crowding (MMC) systems and configurations.
Used for cell separation and organelle isolation.

Other Notes

A nonionic synthetic polymer of sucrose.
Dialyzed
To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Ficoll is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirna González-González et al.
Biotechnology progress, 30(3), 700-707 (2014-01-23)
Aqueous two-phase systems (ATPS) represent a promising strategy for the recovery of CD133(+) stem cells. This particular type of stem cells has great potential for research and clinical applications. Traditional [polyethylene glycol (PEG), dextran (DEX), and ficoll] and novel (Ucon)
Bo Chen et al.
Tissue engineering. Part C, Methods, 19(8), 586-595 (2013-02-21)
Macromolecular crowding (MMC) has been shown to have a beneficial effect on production and maturation of extracellular matrices in a monolayer cell culture model. To explore its potential in tissue engineering, a mixture of Ficoll 70 and Ficoll 400 is
Yanyan Xu et al.
Zhongguo xiu fu chong jian wai ke za zhi = Zhongguo xiufu chongjian waike zazhi = Chinese journal of reparative and reconstructive surgery, 24(4), 406-409 (2010-05-13)
The purity and activity of islets will greatly affect the outcome of xenotransplantation therapy of type 1 diabetes mellitus. To set up an improved method of the isolation and purification of rat islets, which can obtain high-purity, high-yield, and high-viability
Lucie Kosinova et al.
Islets, 11(6), 129-140 (2019-09-10)
Instant Blood-Mediated Inflammatory Reaction (IBMIR) is a major cause of graft loss during pancreatic islet transplantation, leading to a low efficiency of this treatment method and significantly limiting its broader clinical use. Within the procedure, transplanted islets obstruct intrahepatic portal
Frank Brand et al.
Acta neuropathologica, 139(1), 175-192 (2019-09-02)
In search of novel genes associated with glioma pathogenesis, we have previously shown frequent deletions of the KIAA1797/FOCAD gene in malignant gliomas, and a tumor suppressor function of the encoded focadhesin impacting proliferation and migration of glioma cells in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service