Skip to Content
Merck
All Photos(1)

Documents

H17001

Sigma-Aldrich

Wnt3A, human

recombinant, expressed in HEK 293 cells human

Synonym(s):

Wnt, Wnt-3A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in HEK 293 cells human

Assay

>90% (SDS-PAGE)

form

powder

potency

≤20 ng/mL EC50

mol wt

38-42 kDa

impurities

<1 EU/μg endotoxin

storage temp.

−20°C

General description

WNT3A (Wnt family member 3A) gene is mapped to human chromosome 1q42.13. Wnt3A gene is a member of the Wnt gene family that consists of structurally linked genes that encode secreted signaling proteins.

Biochem/physiol Actions

WNT3A (Wnt family member 3A) functions mainly to induce canonical Wnt signaling pathway. Wnt signaling pathway is essential for gastrulation, organogenesis, tissue development, homeostasis and regeneration. Congenital defects, oncogenic transformation and many more disorders results due to abnormal Wnt signaling pathway. During the differentiation of human neuronal progenitor cells, WNT3A (Wnt family member 3A) activates calmodulin-dependent kinase II and increases free calcium in the cytosol via Wnt/Ca2+ signaling pathway. WNT3A is an oncogene and is upregulated in many types of cancer. It serves as a prognostic marker in diagnosing hepatocellular carcinoma. It also promotes metastasis of malignant tumors, including gastric cancer, colorectal cancer, non-small cell lung cancer and breast cancer.

Physical form

Lyophilized from a 0.2 μm filtered solution of 1X PBS pH 7.4 + 500mM NaCl + 1% CHAPS.

Reconstitution

It is recommended to reconstitute the protein to ≤100μg/mL in sterile 1X PBS containing 0.1% endotoxin-free HSA.

Storage and Stability

Upon reconstitution, the cytokine can be stored at 4°C for short term or at -20°C to -80°C in aliquots for long term. Avoid repeated freeze-thaw cycles.

Analysis Note

The activity was determined by dose dependent Alkaline Phosphatase production in MC3T3 E1 mouse preosteoblast cells and stabilization of beta catenin in mouse L929 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Xu et al.
Frontiers in bioengineering and biotechnology, 8, 946-946 (2020-08-28)
Mesenchymal stromal cell senescence and apoptosis have been identified as critical molecular hallmarks in aging. In this study, we used stromal cell sheet culture as an in vitro model to study the progressive changes of cellular senescence, apoptosis and underlying
The WNT target SP5 negatively regulates WNT transcriptional programs in human pluripotent stem cells
Huggins IJ, et al.
Nature Communications, 8(1), 1034-1034 (2017)
The Wnt inhibitor LGK-974 enhances radiosensitivity of HepG2 cells by modulating Nrf2 signaling
Tian D, et al.
International Journal of Oncology, 51(2), 545-554 (2017)
Expression of Wnt3a in hepatocellular carcinoma and its effects on cell cycle and metastasis
Lu C, et al.
International Journal of Oncology, 51(4), 1135-1145 (2017)
Non-canonical pathway induced by Wnt3a regulates ?-catenin via Pyk2 in differentiating human neural progenitor cells.
Narendra Talabattula VA
Biochemical and Biophysical Research Communications, 491(1), 40-46 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service