Skip to Content
Merck
All Photos(6)

Key Documents

359270

Sigma-Aldrich

Sodium tert-butoxide

97%

Synonym(s):

Sodium 2-methylpropan-2-olate, Sodium t-butoxide, Sodium tert-butanolate, Sodium tert-butylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOC(CH3)3
CAS Number:
Molecular Weight:
96.10
Beilstein:
3654215
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.0000026 hPa ( 50 °C)

Assay

97%

form

powder

density

1,025 g/cm3 at 20 °C

SMILES string

[Na+].CC(C)(C)[O-]

InChI

1S/C4H9O.Na/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

MFRIHAYPQRLWNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sodium tert-butoxide may be used as a safe and effective alternative to sodium hydride for the coupling of hydroxyethyl adenine with diethyl p-toluenesulfonyloxymethanephosphonate to form 9-[2-(diethylphosphonomethoxy)ethyl]adenine (diethyl-PMEA), which is a key intermediate to prepare adefovir dipivoxil.[1]
It can also be used:
  • To facilitate the coupling of aryl halides with benzene derivatives in the presence of 10-phenanthroline derivative.[2]
  • To activate first-row transition-metal pre-catalysts.[3]
  • As a base for the amination of aryl chlorides in the presence of a novel palladacyclic precatalyst.[4]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Process optimization in the synthesis of 9-[2-(diethylphosphonomethoxy) ethyl] adenine: replacement of sodium hydride with sodium tert-butoxide as the base for oxygen alkylation.
Yu RH, et al.
Organic Process Research & Development, 3(1), 53-55 (1999)
tert-Butoxide-mediated arylation of benzene with aryl halides in the presence of a catalytic 1, 10-phenanthroline derivative.
Shirakawa E, et al.
Journal of the American Chemical Society, 132(44), 15537-15539 (2010)
Activation and discovery of earth-abundant metal catalysts using sodium tert-butoxide.
Docherty JH, et al.
Nature Chemistry, 9(6), 595-595 (2017)
An air and thermally stable one-component catalyst for the amination of aryl chlorides.
Zim D and Buchwald SL.
Organic Letters, 5(14), 2413-2415 (2003)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service