Skip to Content
Merck
All Photos(6)

Key Documents

156671

Sigma-Aldrich

Potassium tert-butoxide

greener alternative

reagent grade, ≥98%

Synonym(s):

Potassium tert-butylate, Potassium t-butoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COK
CAS Number:
Molecular Weight:
112.21
Beilstein:
3556712
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.22
grade:
reagent grade
form:
solid

grade

reagent grade

Quality Level

vapor pressure

1 mmHg ( 220 °C)

Assay

≥98%

form

solid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

256-258 °C (dec.) (lit.)

greener alternative category

SMILES string

[K+].CC(C)(C)[O-]

InChI

1S/C4H9O.K/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

LPNYRYFBWFDTMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.

Application

Potassium tert-butoxide has been used as a strong base in the enantioselective synthesis of amines by transfer hydrogenation of N-(tertbutylsulfinyl)imines.
It can also be used:
  • To synthesize aliphatic and aromatic amides from corresponding esters and amines.
  • As a base in the intramolecular cyclization of aryl ethers, amines, and amides.
  • As a catalyst to prepare styrene derivatives from aryl halides and alkenes by Mizoroki-Heck reaction.

Used for greener amidation of esters.

tert-Butoxide-Assisted Amidation of Esters under Green Conditions

Potassium tert-butoxide may be used as a base in the intramolecular cyclization of iodo arene to afford benzopyran via microwave method of synthesis.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

tert-Butoxide-assisted amidation of esters under green conditions
Kim Bo Ram, et al.
Synthesis, 44(01), 42-50 (2012)
"Potassium tert-Butoxide Promoted Intramolecular Arylation via a Radical Pathway"
Roman SD, et al.
Organic Letters, 13(12), 3242-3245 (2011)
Mizoroki-Heck-type reaction mediated by potassium tert-butoxide
Shirakawa E, et al.
Angewandte Chemie (International ed. in English), 123(20), 4767-4770 (2011)
Synthesis of highly enantiomerically enriched amines by asymmetric transfer hydrogenation of N-(tert-butylsulfinyl) imines
Guijarro D, et al.
Organic Syntheses (2013)
Wen-Sen He et al.
Bioresource technology, 114, 1-5 (2012-04-03)
The effects of solvents with different log P values, and of lipases on the synthesis of water-soluble plant stanol derivatives were investigated. Results showed that conversion in solvents with log P<0.37 was mainly controlled by the hydrophobicity of the solvent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service